Fast, confident PCR pathogen detection to help protect your business’s brand
Fast, confident PCR pathogen detection to help protect your business’s brand 
Detect foodborne pathogens, and other important food safety and quality targets, quickly and accurately. Empower your operations with rapid reporting to enable faster product release and interventions. Our food safety PCR assays offer qualitative detection of the major pathogens (Salmonella, Listeria, E. coli O157:H7, Cronobacter spp.) and other important targets in food and environmental samples. These PCR food testing solutions are optimized, and in many cases externally validated, for use with our Applied Biosystems™ QuantStudio™ 5 Food Safety Real-Time PCR Instrument on a broad range of food and environmental matrices.
Whether your laboratory is looking to introduce PCR technology for the first time or you want to expand the test services you offer, we can help you find the right solution.
Popular food safety PCR products
PCR food pathogen testing solutions
PCR food pathogen testing solutions
It’s easy to be sure with simple, rapid workflows, and reliable results, run after run
Thermo Scientific™ SureTect™ Real-Time PCR Assays offer few handling steps, simple sample preparation, intuitive easy-to-operate software and universal PCR processing steps. The range covers workflows to detect Salmonella, Listeria, E. coli O157:H7 and Cronobacter in a variety of food and environmental sample types.
salmonella-screening-270x195
Power up Salmonella screening: 3 targets, 1 PCR test
The newest addition to our range of Thermo Scientific PCR food pathogen testing solutions is the Thermo Scientific™ RapidFinder™ Salmonella species, Typhimurium and Enteritidis Multiplex Assay which, like SureTect, follows a simple workflow for simultaneous detection of Salmonella species, Salmonella Typhimurium and Salmonella Enteritidis in poultry, pork and production environment samples.
RapidFinder™
Want to test for an extended range of targets?
Our Applied Biosystems™ MicroSEQ™, RapidFinder™ and TaqMan™ Assays enable you to choose a sample preparation method to suit your needs with workflows incorporating manual and semi-automated options. Maximize throughput with a fast PCR run time of 40 minutes. Validated workflows for detection of Salmonella, Listeria, E. coli O157:H7, non-O157 STEC, STEC and Salmonella Enteritidis are available, as well as TaqMan kits for a broad range of other targets including multiplexed assays for Vibrio, Staphylococcus and Campylobacter.
Which PCR food pathogen tests suit your laboratory's needs?
Food Pathogen PCR Test Selection Guide
Use our selection guide to choose your validated workflow solution by pathogen target and sample type.
View selection guide
Complete flexibility with Applied Biosystems QuantStudio 5 Food Safety Real-Time PCR System
QuantStudio 5 Food Safety Real-Time PCR System
Ensure long-term confidence in your PCR pathogen testing system with the modern, flexible Applied Biosystems™ QuantStudio™ 5 Food Safety Real-Time PCR System. Benefit from the flexibility of a 6-channel, 96-well, cloud-enabled open platform suitable for running our full range of PCR solutions for food safety, authenticity, and quality testing.
  • Efficient: process several hundred samples a day – over 700 tests in a 8h shift
  • Simple: ‘plug and play’ ready to use out of the box
  • Flexible: Open platform suitable for running our full range of PCR solutions of PCR solutions for food safety, authenticity, and quality testing, as well  as various applications including agribusiness and research use. 
  • Cloud-enabled: offering the potential for data sharing, analysis and trending
Used today in thousands of laboratories and with a proven, reliable track record, you can test with confidence and enjoy peace-of-mind knowing your instrument will be fully supported by our own service engineers and dedicated, knowledgeable technical team.
Learn more