Mini Gel Tank specifications
CapacityUp to 2 mini gels
Gel compatibilityGel size: 8 x 8cm
Gel cassette: 10 x 10cm
Thickness: 1.0mm or 1.5mm
Compatible gelsAll Invitrogen precast mini gels, SureCast handcast gels, and Invitrogen empty mini gel cassettes
Buffer chamber requirement400 mL for each mini gel
Optional transfer apparatusMini Blot Module
Transfer capacityUp to 2 mini gels
Compatible power suppliesPowerEase Touch 120W and PowerEase Touch 350W, Owl systems, for other systems such as Hoefer or Bio-Rad use with Novex Power Supply Adapters (Cat. No. ZA10001)
Unit dimensions32 x 11.5 x 16 cm (height with lid)
MaterialPolycarbonate
Electrode wirePlatinum
Electrode limits500 VDC or 100 Watts
Chemical resistanceThe Mini Gel Tank is impervious to most alcohols but not compatible with chlorinated hydrocarbons (e.g., chloroform), aromatic hydrocarbons (e.g., toluene, benzene), or acetone.
Catalog numberA25977
Description
The Mini Gel Tank has a unique tank design that enables convenient side-by-side gel loading and enhanced viewing during use. Each tank can accommodate up to two gels per run where each gel is run in its own separate chamber- making it easy to run just one gel. This advantage allows for lower buffer requirements- only use the buffer needed to run one gel.
The Mini Gel Tank can be used for wet tank transfers using the Mini Blot Module. Two Mini Blot Modules fit into the Mini Gel Tank to allow transfer of two gels at a time. The unique design of the blot module allows for lower transfer buffer- only 220 mL per gel. This lower requirement keeps the amount of methanol needed to a minimum. Learn more about transferring in the Mini Gel Tank here.
Learn how to separate proteins using the Invitrogen Mini Gel Tank.
Video Player is loading.
Current Time 0:00
Duration 5:24
Loaded: 0.00%
Stream Type LIVE
Remaining Time 5:24
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Learn how to perform a western wet tank transfer using the Invitrogen Mini Blot Module in the Mini Gel Tank.
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
        Recommended electrophoresis run conditions
        Gel TypeVoltage (V)Current (mA)Ending CurrentRun Time
        Bolt Bis-Tris Plus 4-12% (MES)2001607020
        Bolt Bis-Tris Plus 4-12% (MOPS)2001605035
        NuPAGE 4-12% BisTris (MES)2001609030
        NuPAGE 4-12% BisTris (MOPS)2001405042
        Novex 4-20% TrisGlycine (denatured)1254010100
        Novex 4-20% TrisGlycine (native)125301090
        NuPAGE 3-8% TrisAcetate (denatured)150602050
        NuPAGE 3-8% TrisAcetate (native)1504010100
        Novex 10-20% Tricine1251104065
        NativePAGE 3-12%15010<1080
        pH 3-10 IEFprogram85150
        10% Zymogram (Gelatin)125401090
        10% TBE-Urea (DNA)180101050
        Catalog # Name Size Price (USD) Qty
        A25977 Mini Gel Tank Each
        735.65
        Online Exclusive
        760.00
        Save 24.35 (3%)
        A25944 Mini Gel Tank Lid Each
        211.65
        Online Exclusive
        218.00
        Save 6.35 (3%)
        A25945 Cassette Clamp, right Each
        173.65
        Online Exclusive
        179.00
        Save 5.35 (3%)
        A25946 Cassette Clamp, left Each
        170.65
        Online Exclusive
        176.00
        Save 5.35 (3%)
        A25950 Mini Gel Tank Base Each
        43.65
        Online Exclusive
        44.64
        NW2000 Mini Gel Tank and Blot Module Set Each
        1,314.65
        Online Exclusive
        1,368.00
        Save 53.35 (4%)
        B4478641 Gel Runner Tank Each
        197.00
        Mini Gel Tank specifications
        CapacityUp to 2 mini gels
        Gel compatibilityGel size: 8 x 8cm
        Gel cassette: 10 x 10cm
        Thickness: 1.0mm or 1.5mm
        Compatible gelsAll Invitrogen precast mini gels, SureCast handcast gels, and Invitrogen empty mini gel cassettes
        Buffer chamber requirement400 mL for each mini gel
        Optional transfer apparatusMini Blot Module
        Transfer capacityUp to 2 mini gels
        Compatible power suppliesPowerEase Touch 120W and PowerEase Touch 350W, Owl systems, for other systems such as Hoefer or Bio-Rad use with Novex Power Supply Adapters (Cat. No. ZA10001)
        Unit dimensions32 x 11.5 x 16 cm (height with lid)
        MaterialPolycarbonate
        Electrode wirePlatinum
        Electrode limits500 VDC or 100 Watts
        Chemical resistanceThe Mini Gel Tank is impervious to most alcohols but not compatible with chlorinated hydrocarbons (e.g., chloroform), aromatic hydrocarbons (e.g., toluene, benzene), or acetone.
        Catalog numberA25977
        Description
        The Mini Gel Tank has a unique tank design that enables convenient side-by-side gel loading and enhanced viewing during use. Each tank can accommodate up to two gels per run where each gel is run in its own separate chamber- making it easy to run just one gel. This advantage allows for lower buffer requirements- only use the buffer needed to run one gel.
        The Mini Gel Tank can be used for wet tank transfers using the Mini Blot Module. Two Mini Blot Modules fit into the Mini Gel Tank to allow transfer of two gels at a time. The unique design of the blot module allows for lower transfer buffer- only 220 mL per gel. This lower requirement keeps the amount of methanol needed to a minimum. Learn more about transferring in the Mini Gel Tank here.
        Learn how to separate proteins using the Invitrogen Mini Gel Tank.
        Video Player is loading.
        Current Time 0:00
        Duration 0:00
        Loaded: 0%
        Stream Type LIVE
        Remaining Time 0:00
        1x
          • Chapters
          • descriptions off, selected
          • captions off, selected
          • en (Main), selected
          Learn how to perform a western wet tank transfer using the Invitrogen Mini Blot Module in the Mini Gel Tank.
          Video Player is loading.
          Current Time 0:00
          Duration 0:00
          Loaded: 0%
          Stream Type LIVE
          Remaining Time 0:00
          1x
            • Chapters
            • descriptions off, selected
            • captions off, selected
              Recommended electrophoresis run conditions
              Gel TypeVoltage (V)Current (mA)Ending CurrentRun Time
              Bolt Bis-Tris Plus 4-12% (MES)2001607020
              Bolt Bis-Tris Plus 4-12% (MOPS)2001605035
              NuPAGE 4-12% BisTris (MES)2001609030
              NuPAGE 4-12% BisTris (MOPS)2001405042
              Novex 4-20% TrisGlycine (denatured)1254010100
              Novex 4-20% TrisGlycine (native)125301090
              NuPAGE 3-8% TrisAcetate (denatured)150602050
              NuPAGE 3-8% TrisAcetate (native)1504010100
              Novex 10-20% Tricine1251104065
              NativePAGE 3-12%15010<1080
              pH 3-10 IEFprogram85150
              10% Zymogram (Gelatin)125401090
              10% TBE-Urea (DNA)180101050
              Catalog # Name Size Price (USD) Qty
              A25977 Mini Gel Tank Each
              735.65
              Online Exclusive
              760.00
              Save 24.35 (3%)
              A25944 Mini Gel Tank Lid Each
              211.65
              Online Exclusive
              218.00
              Save 6.35 (3%)
              A25945 Cassette Clamp, right Each
              173.65
              Online Exclusive
              179.00
              Save 5.35 (3%)
              A25946 Cassette Clamp, left Each
              170.65
              Online Exclusive
              176.00
              Save 5.35 (3%)
              A25950 Mini Gel Tank Base Each
              43.65
              Online Exclusive
              44.64
              NW2000 Mini Gel Tank and Blot Module Set Each
              1,314.65
              Online Exclusive
              1,368.00
              Save 53.35 (4%)
              B4478641 Gel Runner Tank Each
              197.00
              For Research Use Only. Not for use in diagnostic procedures.