Capillary endothelial cells and microglia in a rat brain cryosection.
Capillary endothelial cells and microglia in a rat brain cryosection labeled with the green-fluorescent Alexa Fluor® 488 isolectin IB4 (Cat. No. I21411). Neurons were identified using the NeuroTrace® 530/615 red-fluorescent Nissl stain (Cat. No. N21482) and nuclei were counterstained with blue-fluorescent DAPI (Cat. No. D1306, D3571, D21490). The digital image was obtained using filters appropriate for fluorescein, rhodamine, and DAPI.
Capillary endothelial cells and microglia in a rat brain cryosection labeled with the green-fluorescent Alexa Fluor® 488 isolectin IB<sub>4</sub> (Cat. No. I21411). Neurons were identified using the NeuroTrace® 530/615 red-fluorescent Nissl stain (Cat. No. N21482) and nuclei were counterstained with blue-fluorescent DAPI (Cat. No. D1306, D3571, D21490). The digital image was obtained using filters appropriate for fluorescein, rhodamine, and DAPI.
Related Products
Catalog # Name Size Price (USD) Qty
I21411 Isolectin GS-IB4 From Griffonia simplicifolia, Alexa Fluor™ 488 Conjugate Each
544.65
Online Exclusive
562.00
Save 17.35 (3%)
D1306 DAPI (4',6-diamidino-2-phenylindole, dihydrochloride), 10 mg, 10 mg Each
152.65
Online Exclusive
156.00
Save 3.35 (2%)
Related Images