Invitrogen™
eBioscience™ Fixation/Permeabilization Concentrate
Catalog number: 00-5123-43
Have Questions?
Invitrogen™
eBioscience™ Fixation/Permeabilization Concentrate
Catalog number: 00-5123-43
Have Questions?
Catalog Number
00-5123-43
Unit Size
30 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
00-5123-4330 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Fixation/Permeabilization Concentrate is intended to be used with antibodies to transcription factors and nuclear proteins, such as Foxp3 and Ki-67, as well as cytokines and chemokines. This must be used in combination with the Fixation/Permeabilization Diluent (cat. 00-5223) and the Permeabilization Buffer (10X) (cat. 00-8333). Alternatively, all buffers required for staining with these antibodies can be purchased as a set in the Foxp3 Transcription Factor Staining Buffer Set (cat. 00-5523).
Reported Application
Intracellular Staining Followed by Flow Cytometric Analysis
WARNING: Cancer – www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Includes
One 30 mL bottle
Quantity
30 mL
Reagent Type
Fixation Reagent, Permeabilization Reagent
Product Line
eBioscience™
Product Type
Fixation/Permeabilization Buffer