Informational:This product is equivalent to C36950 - CountBright™ Absolute Counting Beads.
Invitrogen™
123count eBeads™ Counting Beads
Catalog number: 01-1234-42
Have Questions?
Invitrogen™
123count eBeads™ Counting Beads
Catalog number: 01-1234-42
Have Questions?
Catalog Number
01-1234-42
Unit Size
100 tests
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
01-1234-42100 tests
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
123count eBeads are intended for use in absolute counting of cells or other particles by flow cytometry. These are 7 μm microparticles containing encapsulated dyes compatible with blue (488 nm) and violet (405 nm) excitation sources and emitting fluorescence between approximately 500 nm and 750 nm. They are supplied at a known concentration in 0.05% Tween-20 with 2 mM sodium azide. Collecting carefully suspended and measured quantities of cell:bead mixtures on a flow cytometer makes the ratiometric enumeration of cells possible.
Reported Application
Flow Cytometric Analysis
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)
Flow Cytometer
Product Line
eBeads™
Product Type
Cell Counting Bead