Invitrogen™

UltraComp eBeads™ Compensation Beads

Catalog number: 01-2222-42
Have Questions?
Invitrogen™

UltraComp eBeads™ Compensation Beads

Catalog number: 01-2222-42
Have Questions?
Catalog Number
01-2222-42
Unit Size
100 tests
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
25 tests
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
01-2222-42100 tests
-
01-2222-4125 tests
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
UltraComp eBeads™ react with antibodies of mouse, rat, and hamster origin, and are immunoglobulin light chain-independent. They are designed for use in compensation with all fluorochromes excited by ultraviolet (355 nm) violet (405 nm), blue (488 nm), green (532 nm), yellow-green (561 nm), and red (633-640 nm) lasers. The beads are spherical particles that can be stained with individual fluorochrome-conjugated antibodies for use as single-color compensation controls.

Each drop of beads contains two populations: a positive population that will capture any mouse, rat, or hamster antibody and a negative population that will not react with antibody. When a fluorochrome-conjugated antibody is added to the beads, both positive and negative populations result. This bimodal distribution can be used for single-color compensation controls in multicolor flow cytometry experiments.

UltraComp eBeads are compatible with all fluorochromes excited by an ultraviolet (355 nm) or violet (405 nm) laser. They should be used with standard staining buffers which contain PBS or HBSS, protein such as BSA or FBS, and sodium azide. No other additives should be used. For more information, please contact technical support.

Reactivity/Species
Hamster, Mouse, Rat

Reported Application
Flow Cytometric Analysis

For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment)
Flow Cytometer
No. of Tests
100 Tests
Product Line
eBeads™
Product Type
Compensation Beads

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text