Thermo Scientific™

Dionex™ Carbonate, Methanesulfonic Acid, and Hydroxide EGC Eluent Generator Cartridges

Catalog number: 072076
Have Questions?
Thermo Scientific™

Dionex™ Carbonate, Methanesulfonic Acid, and Hydroxide EGC Eluent Generator Cartridges

Catalog number: 072076
Have Questions?

Achieve reproducible and reliable chromatography separations using eluent generation. Thermo Scientific™ Dionex™ EGC Eluent Generator Cartridges provide consistent, reproducible eluent generation, ensuring confidence in your results. These cartridges eliminate the manual work of eluent preparation and allow you to focus on the science. Eluent generation also removes the need to handle the acids and bases traditionally required for manual IC eluent preparation, saving cost on chemical storage and disposal.

 
Catalog Number
072076
Unit Size
Each
Description
Dionex EGC-KOH Potassium Hydroxide Eluent Generator Cartridge, Capillary
For Use With (Equipment)
ICS-4000 Capillary HPIC System and Dionex ICS-5000+ Capillary System
Solvents
25% methanol
Price (USD)
Full specifications
Quantity
For Use With (Equipment)ICS-4000 Capillary HPIC System and Dionex ICS-5000+ Capillary System
TypePotassium Hydroxide Eluent Generator Cartridge
Flow Rate1 to 30 μL/min. (0.001 to 0.030 mL/min.)
PressureMaximum: 34.5 MPa (5000 psi)
Solvents25% methanol
OperationDisposable means of producing potassium hydroxide (KOH) eluent for anion separations.
Concentration0.1 to 200 mM
DescriptionDionex EGC-KOH Potassium Hydroxide Eluent Generator Cartridge, Capillary
Unit SizeEach
Catalog Number
Unit Size
Each 1
Description
Dionex EGC 500 K2CO3 Potassium Carbonate Eluent Generator Cartridge
For Use With (Equipment)
Dionex ICS-5000+, ICS-6000 HPIC Systems
Solvents
No solvents
Price (USD)
Full specifications
Quantity
For Use With (Equipment)Dionex ICS-5000+, ICS-6000 HPIC Systems
TypePotassium Carbonate Eluent Generator Cartridge
Flow Rate0.1 to 3.00 mL/min.
Pressure34.5 MPa (3000 psi)
SolventsNo solvents
OperationGenerates carbonate-only eluent as potassium carbonate
Concentration0.1 to 100 mM
Required OptionsEPM and EGC Carbonate Mixer Kit
DescriptionDionex EGC 500 K2CO3 Potassium Carbonate Eluent Generator Cartridge
Unit SizeEach 1
Catalog Number
Unit Size
Each
Description
Dionex EGC III KOH Potassium Hydroxide Eluent Generator Cartridge
For Use With (Equipment)
Traditional RFIC-EG Systems up to 3000 psi
Solvents
25% methanol
Price (USD)
Full specifications
Quantity
For Use With (Equipment)Traditional RFIC-EG Systems up to 3000 psi
TypePotassium Hydroxide Eluent Generator Cartridge
Flow Rate0.1 to 3.00 mL/min.
PressureMaximum: 21 MPa (3000 psi)
Solvents25% methanol
OperationFor the production of hydroxide eluents.
Concentration0.1 to 100 mM
DescriptionDionex EGC III KOH Potassium Hydroxide Eluent Generator Cartridge
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Dionex EGC III NaOH Sodium Hydroxide Eluent Generator Cartridge
For Use With (Equipment)
Traditional RFIC-EG Systems up to 3000 psi
Solvents
25% methanol
Price (USD)
Full specifications
Quantity
For Use With (Equipment)Traditional RFIC-EG Systems up to 3000 psi
TypeSodium Hydroxide Eluent Generator Cartridge
Flow Rate0.1 to 3.00 mL/min.
PressureMaximum: 21 MPa (3000 psi)
Solvents25% methanol
OperationFor the production of hydroxide eluents.
Concentration0.1 to 100 mM
DescriptionDionex EGC III NaOH Sodium Hydroxide Eluent Generator Cartridge
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Dionex EGC III LiOH Lithium Hydroxide Eluent Generator Cartridge
For Use With (Equipment)
Traditional RFIC-EG Systems up to 3000 psi
Solvents
25% methanol
Price (USD)
Full specifications
Quantity
For Use With (Equipment)Traditional RFIC-EG Systems up to 3000 psi
TypeLithium Hydroxide Eluent Generator Cartridge
Flow Rate0.1 to 3.00 mL/min.
PressureMaximum: 21 MPa (3000 psi)
Solvents25% methanol
OperationFor the production of hydroxide eluents.
Concentration0.1 to 80 mM
DescriptionDionex EGC III LiOH Lithium Hydroxide Eluent Generator Cartridge
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Dionex EGC III MSA Methanesulfonic Acid Eluent Generator Cartridge
For Use With (Equipment)
Traditional RFIC-EG Systems up to 3000 psi
Solvents
No solvents
Price (USD)
Full specifications
Quantity
For Use With (Equipment)Traditional RFIC-EG Systems up to 3000 psi
TypeMethanesulfonic Acid Eluent Generator Cartridge
Flow Rate0.1 to 3.00 mL/min.
PressureMaximum: 21 MPa (3000 psi)
SolventsNo solvents
OperationDisposable means of producing eluent for cation separations.
Concentration0.1 to 100 mM
DescriptionDionex EGC III MSA Methanesulfonic Acid Eluent Generator Cartridge
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Dionex EGC-MSA Methanesulfonic Acid Eluent Generator Cartridge, Capillary
For Use With (Equipment)
ICS-4000 Capillary HPIC System and Dionex ICS-5000+ Capillary System
Solvents
No solvents
Price (USD)
Full specifications
Quantity
For Use With (Equipment)ICS-4000 Capillary HPIC System and Dionex ICS-5000+ Capillary System
TypeMethanesulfonic Acid Eluent Generator Cartridge
Flow Rate1 to 30 μL/min. (0.001 to 0.030 mL/min.)
PressureMaximum: 34.5 MPa (5000 psi)
SolventsNo solvents
OperationFor the production methanesulfonic acid eluents.
Concentration0.1 to 200 mM
DescriptionDionex EGC-MSA Methanesulfonic Acid Eluent Generator Cartridge, Capillary
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Dionex EGC 500 KOH Potassium Hydroxide Eluent Generator Cartridge
For Use With (Equipment)
Integrion, ICS-5000+, ICS-6000 HPIC Systems
Solvents
EGC 500 KOH: 25% methanol, EGC 500 MSA: no solvents
Price (USD)
Full specifications
Quantity
For Use With (Equipment)Integrion, ICS-5000+, ICS-6000 HPIC Systems
TypePotassium Hydroxide Eluent Generator Cartridge
Flow Rate0.01 to 3.00 mL/min.
PressureMaximum: 34.5 MPa (5000 psi)
SolventsEGC 500 KOH: 25% methanol, EGC 500 MSA: no solvents
Concentration0.1 to 100 mM
DescriptionDionex EGC 500 KOH Potassium Hydroxide Eluent Generator Cartridge
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Dionex EGC 500 MSA Methanesulfonic Acid Eluent Generator Cartridge
For Use With (Equipment)
Integrion, ICS-5000+, ICS-6000 HPIC Systems
Solvents
No solvents
Price (USD)
Full specifications
Quantity
For Use With (Equipment)Integrion, ICS-5000+, ICS-6000 HPIC Systems
TypeMethanesulfonic Acid Eluent Generator Cartridge
Flow Rate0.10 to 3.00 mL/min.
PressureMaximum: 34.5 MPa (5000 psi)
SolventsNo solvents
Concentration0.1 to 100 mM
DescriptionDionex EGC 500 MSA Methanesulfonic Acid Eluent Generator Cartridge
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Dionex EGC 400 KOH Potassium Hydroxide Eluent Generator Cartridge
For Use With (Equipment)
Dual EGC Mode on Dionex ICS-6000 HPIC Standard/Microbore or Hybrid Systems
Solvents
No solvents recommended
Price (USD)
Full specifications
Quantity
For Use With (Equipment)Dual EGC Mode on Dionex ICS-6000 HPIC Standard/Microbore or Hybrid Systems
TypePotassium Hydroxide Eluent Generator Cartridge
Flow Rate20 to 200 μL/min.
PressureMaximum: 34.5 MPa (5000 psi)
SolventsNo solvents recommended
OperationDual EGC operating mode only.
Concentration0.1 to 200 mM
DescriptionDionex EGC 400 KOH Potassium Hydroxide Eluent Generator Cartridge
Unit SizeEach
Showing 10 of 11Show more
Catalog NumberSpecificationsUnit SizeDescriptionFor Use With (Equipment)SolventsPrice (USD)Quantity
072076Full specifications
EachDionex EGC-KOH Potassium Hydroxide Eluent Generator Cartridge, CapillaryICS-4000 Capillary HPIC System and Dionex ICS-5000+ Capillary System25% methanol
For Use With (Equipment)ICS-4000 Capillary HPIC System and Dionex ICS-5000+ Capillary System
TypePotassium Hydroxide Eluent Generator Cartridge
Flow Rate1 to 30 μL/min. (0.001 to 0.030 mL/min.)
PressureMaximum: 34.5 MPa (5000 psi)
Solvents25% methanol
OperationDisposable means of producing potassium hydroxide (KOH) eluent for anion separations.
Concentration0.1 to 200 mM
DescriptionDionex EGC-KOH Potassium Hydroxide Eluent Generator Cartridge, Capillary
Unit SizeEach
088453Full specifications
Each 1Dionex EGC 500 K2CO3 Potassium Carbonate Eluent Generator CartridgeDionex ICS-5000+, ICS-6000 HPIC SystemsNo solvents
For Use With (Equipment)Dionex ICS-5000+, ICS-6000 HPIC Systems
TypePotassium Carbonate Eluent Generator Cartridge
Flow Rate0.1 to 3.00 mL/min.
Pressure34.5 MPa (3000 psi)
SolventsNo solvents
OperationGenerates carbonate-only eluent as potassium carbonate
Concentration0.1 to 100 mM
Required OptionsEPM and EGC Carbonate Mixer Kit
DescriptionDionex EGC 500 K2CO3 Potassium Carbonate Eluent Generator Cartridge
Unit SizeEach 1
074532Full specifications
EachDionex EGC III KOH Potassium Hydroxide Eluent Generator CartridgeTraditional RFIC-EG Systems up to 3000 psi25% methanol
For Use With (Equipment)Traditional RFIC-EG Systems up to 3000 psi
TypePotassium Hydroxide Eluent Generator Cartridge
Flow Rate0.1 to 3.00 mL/min.
PressureMaximum: 21 MPa (3000 psi)
Solvents25% methanol
OperationFor the production of hydroxide eluents.
Concentration0.1 to 100 mM
DescriptionDionex EGC III KOH Potassium Hydroxide Eluent Generator Cartridge
Unit SizeEach
074533Full specifications
EachDionex EGC III NaOH Sodium Hydroxide Eluent Generator CartridgeTraditional RFIC-EG Systems up to 3000 psi25% methanol
For Use With (Equipment)Traditional RFIC-EG Systems up to 3000 psi
TypeSodium Hydroxide Eluent Generator Cartridge
Flow Rate0.1 to 3.00 mL/min.
PressureMaximum: 21 MPa (3000 psi)
Solvents25% methanol
OperationFor the production of hydroxide eluents.
Concentration0.1 to 100 mM
DescriptionDionex EGC III NaOH Sodium Hydroxide Eluent Generator Cartridge
Unit SizeEach
074534Full specifications
EachDionex EGC III LiOH Lithium Hydroxide Eluent Generator CartridgeTraditional RFIC-EG Systems up to 3000 psi25% methanol
For Use With (Equipment)Traditional RFIC-EG Systems up to 3000 psi
TypeLithium Hydroxide Eluent Generator Cartridge
Flow Rate0.1 to 3.00 mL/min.
PressureMaximum: 21 MPa (3000 psi)
Solvents25% methanol
OperationFor the production of hydroxide eluents.
Concentration0.1 to 80 mM
DescriptionDionex EGC III LiOH Lithium Hydroxide Eluent Generator Cartridge
Unit SizeEach
074535Full specifications
EachDionex EGC III MSA Methanesulfonic Acid Eluent Generator CartridgeTraditional RFIC-EG Systems up to 3000 psiNo solvents
For Use With (Equipment)Traditional RFIC-EG Systems up to 3000 psi
TypeMethanesulfonic Acid Eluent Generator Cartridge
Flow Rate0.1 to 3.00 mL/min.
PressureMaximum: 21 MPa (3000 psi)
SolventsNo solvents
OperationDisposable means of producing eluent for cation separations.
Concentration0.1 to 100 mM
DescriptionDionex EGC III MSA Methanesulfonic Acid Eluent Generator Cartridge
Unit SizeEach
072077Full specifications
EachDionex EGC-MSA Methanesulfonic Acid Eluent Generator Cartridge, CapillaryICS-4000 Capillary HPIC System and Dionex ICS-5000+ Capillary SystemNo solvents
For Use With (Equipment)ICS-4000 Capillary HPIC System and Dionex ICS-5000+ Capillary System
TypeMethanesulfonic Acid Eluent Generator Cartridge
Flow Rate1 to 30 μL/min. (0.001 to 0.030 mL/min.)
PressureMaximum: 34.5 MPa (5000 psi)
SolventsNo solvents
OperationFor the production methanesulfonic acid eluents.
Concentration0.1 to 200 mM
DescriptionDionex EGC-MSA Methanesulfonic Acid Eluent Generator Cartridge, Capillary
Unit SizeEach
075778Full specifications
EachDionex EGC 500 KOH Potassium Hydroxide Eluent Generator CartridgeIntegrion, ICS-5000+, ICS-6000 HPIC SystemsEGC 500 KOH: 25% methanol, EGC 500 MSA: no solvents
For Use With (Equipment)Integrion, ICS-5000+, ICS-6000 HPIC Systems
TypePotassium Hydroxide Eluent Generator Cartridge
Flow Rate0.01 to 3.00 mL/min.
PressureMaximum: 34.5 MPa (5000 psi)
SolventsEGC 500 KOH: 25% methanol, EGC 500 MSA: no solvents
Concentration0.1 to 100 mM
DescriptionDionex EGC 500 KOH Potassium Hydroxide Eluent Generator Cartridge
Unit SizeEach
075779Full specifications
EachDionex EGC 500 MSA Methanesulfonic Acid Eluent Generator CartridgeIntegrion, ICS-5000+, ICS-6000 HPIC SystemsNo solvents
For Use With (Equipment)Integrion, ICS-5000+, ICS-6000 HPIC Systems
TypeMethanesulfonic Acid Eluent Generator Cartridge
Flow Rate0.10 to 3.00 mL/min.
PressureMaximum: 34.5 MPa (5000 psi)
SolventsNo solvents
Concentration0.1 to 100 mM
DescriptionDionex EGC 500 MSA Methanesulfonic Acid Eluent Generator Cartridge
Unit SizeEach
302766Full specifications
EachDionex EGC 400 KOH Potassium Hydroxide Eluent Generator CartridgeDual EGC Mode on Dionex ICS-6000 HPIC Standard/Microbore or Hybrid SystemsNo solvents recommended
For Use With (Equipment)Dual EGC Mode on Dionex ICS-6000 HPIC Standard/Microbore or Hybrid Systems
TypePotassium Hydroxide Eluent Generator Cartridge
Flow Rate20 to 200 μL/min.
PressureMaximum: 34.5 MPa (5000 psi)
SolventsNo solvents recommended
OperationDual EGC operating mode only.
Concentration0.1 to 200 mM
DescriptionDionex EGC 400 KOH Potassium Hydroxide Eluent Generator Cartridge
Unit SizeEach
Showing 10 of 11Show more

How Dionex Eluent Generator Cartridges improve IC analysis:

  • Obtain outstanding run-to-run reproducibility by eliminating variability from manual eluent preparation
  • Increase confidence in results by eliminating errors from manual eluent preparation
  • Support large application range
  • Increase workflow efficiency by automating eluent preparation
  • Improve operator's safety by reducing exposure to hazardous chemicals
  • Reduce system maintenance since pump is only exposed to deionized water

Product features:

  • Automatic production of high-purity eluents
  • Support broad range of eluent concentrations
  • Support broad range of flow rates
  • Support pressure up to 5,000 psi
  • Enable isocratic as well as gradient separations
  • Dual EGC mode on Thermo Scientific™ Dionex™ ICS-6000 HPIC System specifically designed for complex carbohydrate analysis (Thermo Scientific™ Dionex™ EGC 400 cartridges)
  • Closed system keeping chemicals contained and isolated

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for six (6) months thereafter.

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text