Achieve reproducible and reliable chromatography separations using eluent generation. Thermo Scientific™ Dionex™ EGC Eluent Generator Cartridges provide consistent, reproducible eluent generation, ensuring confidence in your results. These cartridges eliminate the manual work of eluent preparation and allow you to focus on the science. Eluent generation also removes the need to handle the acids and bases traditionally required for manual IC eluent preparation, saving cost on chemical storage and disposal.
Catalog Number
074533
Unit Size
Each
Description
Dionex EGC III NaOH Sodium Hydroxide Eluent Generator Cartridge
For Use With (Equipment)
Traditional RFIC-EG Systems up to 3000 psi
Solvents
25% methanol
Price (USD)
1,860.00
Full specifications
Quantity
For Use With (Equipment)
Traditional RFIC-EG Systems up to 3000 psi
Type
Sodium Hydroxide Eluent Generator Cartridge
Flow Rate
0.1 to 3.00 mL/min.
Pressure
Maximum: 21 MPa (3000 psi)
Solvents
25% methanol
Operation
For the production of hydroxide eluents.
Concentration
0.1 to 100 mM
Description
Dionex EGC III NaOH Sodium Hydroxide Eluent Generator Cartridge
How Dionex Eluent Generator Cartridges improve IC analysis:
Obtain outstanding run-to-run reproducibility by eliminating variability from manual eluent preparation
Increase confidence in results by eliminating errors from manual eluent preparation
Support large application range
Increase workflow efficiency by automating eluent preparation
Improve operator's safety by reducing exposure to hazardous chemicals
Reduce system maintenance since pump is only exposed to deionized water
Product features:
Automatic production of high-purity eluents
Support broad range of eluent concentrations
Support broad range of flow rates
Support pressure up to 5,000 psi
Enable isocratic as well as gradient separations
Dual EGC mode on Thermo Scientific™ Dionex™ ICS-6000 HPIC System specifically designed for complex carbohydrate analysis (Thermo Scientific™ Dionex™ EGC 400 cartridges)
Closed system keeping chemicals contained and isolated
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for six (6) months thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.