Thermo Scientific™

MAbPac™ SEC-1 Size Exclusion Chromatography HPLC Columns

Catalog number: 075592
Have Questions?
Thermo Scientific™

MAbPac™ SEC-1 Size Exclusion Chromatography HPLC Columns

Catalog number: 075592
Have Questions?

Use Thermo Scientific™ MAbPac™ SEC-1 Size Exclusion Chromatography HPLC Columns for high-resolution separation of monoclonal antibody (mAb) analysis, including the monomers, aggregates and Fab and Fc fragments resulting from proteolysis. These size-exclusion chromatography (SEC) columns provide superior performance, even under nondenaturing conditions, and in high- and low-salt mobile phases and volatile eluents.

 
Catalog Number
075592
Unit Size
Each
Particle Size
5 μm
Diameter (Metric)
4 mm
Length (Metric)
150 mm
Column Type
Size Exclusion
Price (USD)
Full specifications
Quantity
Column ChemistryProprietary Diol
Column FormatAnalytical Column
Diameter (Metric)4 mm
Length (Metric)150 mm
Max. Pressure600 psi (40 bar)
Particle Size5 μm
pH2.5 to 7.5
Pore Size300 Å
Solvents100% Organic Solvents
Temperature30°C
USP TypeL59
Stationary PhaseDiol (OH)
Product LineMAbPac
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Column TypeSize Exclusion
PhaseSize Exclusion
MaterialPEEK
TechniqueBio-LC
Unit SizeEach
Catalog Number
Unit Size
Each
Particle Size
5 μm
Diameter (Metric)
7.8 mm
Length (Metric)
300 mm
Column Type
Size Exclusion
Price (USD)
Full specifications
Quantity
Column ChemistryProprietary Diol
Column FormatAnalytical Column
Diameter (Metric)7.8 mm
Flow Rate760 to 1000 μL/min
For Use WithProtein Mol wt Range 10 to 1000 kDa
Length (Metric)300 mm
Max. Pressure1000 psi (70 bar)
Particle Size5 μm
pH2.5 to 7.5
Pore Size300 Å
Solvents100% Organic Solvents
Temperature30°C
Stationary PhaseDiol (OH)
Product LineMAbPac
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Column TypeSize Exclusion
PhaseSize Exclusion
TechniqueBio-LC
Unit SizeEach
Catalog Number
Unit Size
Each
Particle Size
5 μm
Diameter (Metric)
4 mm
Length (Metric)
300 mm
Column Type
Size Exclusion
Price (USD)
Full specifications
Quantity
Column ChemistryProprietary Diol
Column FormatAnalytical Column
Diameter (Metric)4 mm
Length (Metric)300 mm
Max. Pressure1000 psi (70 bar)
Particle Size5 μm
pH2.5 to 7.5
Pore Size300 Å
Solvents100% Organic Solvents
Temperature30°C
USP TypeL59
Stationary PhaseDiol (OH)
Product LineMAbPac
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Column TypeSize Exclusion
PhaseSize Exclusion
MaterialPEEK
TechniqueBio-LC
Unit SizeEach
Catalog Number
Unit Size
Each
Particle Size
5 μm
Diameter (Metric)
4 mm
Length (Metric)
50 mm
Column Type
Size Exclusion
Price (USD)
Full specifications
Quantity
Column ChemistryProprietary Diol
Column FormatGuard Column
Diameter (Metric)4 mm
Length (Metric)50 mm
Max. Pressure[Not Rated]
Particle Size5 μm
pH2.5 to 7.5
Pore Size300 Å
Solvents100% Organic Solvents
Temperature30°C
USP TypeL59
Stationary PhaseDiol (OH)
Product LineMAbPac
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Column TypeSize Exclusion
PhaseSize Exclusion
MaterialPEEK
TechniqueBio-LC
Unit SizeEach
Catalog Number
088790
also known as 88790
Unit Size
Each
Particle Size
5 μm
Diameter (Metric)
2.1 mm
Length (Metric)
150 mm
Column Type
Size Exclusion
Price (USD)
Full specifications
Quantity
-
Column ChemistrySize Exclusion
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
Length (Metric)150 mm
Max. Pressure600 psi (40 bar)
Particle Size5 μm
pH2.5 to 7.5
Pore Size300 Å
Solvents100% Organic Solvents
Temperature30°C
USP TypeL59
Stationary PhaseProprietary Diol
Product LineMAbPac
Packing MaterialSpherical, High Purity, Porous Silica
Column TypeSize Exclusion
PhaseSize Exclusion
MaterialStainless Steel
TechniqueBio-LC
Unit SizeEach
Catalog Number
Unit Size
Each
Particle Size
5 μm
Diameter (Metric)
2.1 mm
Length (Metric)
300 mm
Column Type
Size Exclusion
Price (USD)
Full specifications
Quantity
-
Column ChemistrySize Exclusion
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
Length (Metric)300 mm
Max. Pressure1000 psi (70 bar)
Particle Size5 μm
pH2.5 to 7.5
Pore Size300 Å
Solvents100% Organic Solvents
Temperature30°C
USP TypeL59
Stationary PhaseProprietary Diol
Product LineMAbPac
Packing MaterialSpherical, High Purity, Porous Silica
Column TypeSize Exclusion
PhaseSize Exclusion
MaterialStainless Steel
Unit SizeEach
Showing 6 of 6
Catalog NumberSpecificationsUnit SizeParticle SizeDiameter (Metric)Length (Metric)Column TypePrice (USD)Quantity
075592Full specifications
Each5 μm4 mm150 mmSize Exclusion
Column ChemistryProprietary Diol
Column FormatAnalytical Column
Diameter (Metric)4 mm
Length (Metric)150 mm
Max. Pressure600 psi (40 bar)
Particle Size5 μm
pH2.5 to 7.5
Pore Size300 Å
Solvents100% Organic Solvents
Temperature30°C
USP TypeL59
Stationary PhaseDiol (OH)
Product LineMAbPac
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Column TypeSize Exclusion
PhaseSize Exclusion
MaterialPEEK
TechniqueBio-LC
Unit SizeEach
088460Full specifications
Each5 μm7.8 mm300 mmSize Exclusion
Column ChemistryProprietary Diol
Column FormatAnalytical Column
Diameter (Metric)7.8 mm
Flow Rate760 to 1000 μL/min
For Use WithProtein Mol wt Range 10 to 1000 kDa
Length (Metric)300 mm
Max. Pressure1000 psi (70 bar)
Particle Size5 μm
pH2.5 to 7.5
Pore Size300 Å
Solvents100% Organic Solvents
Temperature30°C
Stationary PhaseDiol (OH)
Product LineMAbPac
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Column TypeSize Exclusion
PhaseSize Exclusion
TechniqueBio-LC
Unit SizeEach
074696Full specifications
Each5 μm4 mm300 mmSize Exclusion
Column ChemistryProprietary Diol
Column FormatAnalytical Column
Diameter (Metric)4 mm
Length (Metric)300 mm
Max. Pressure1000 psi (70 bar)
Particle Size5 μm
pH2.5 to 7.5
Pore Size300 Å
Solvents100% Organic Solvents
Temperature30°C
USP TypeL59
Stationary PhaseDiol (OH)
Product LineMAbPac
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Column TypeSize Exclusion
PhaseSize Exclusion
MaterialPEEK
TechniqueBio-LC
Unit SizeEach
074697Full specifications
Each5 μm4 mm50 mmSize Exclusion
Column ChemistryProprietary Diol
Column FormatGuard Column
Diameter (Metric)4 mm
Length (Metric)50 mm
Max. Pressure[Not Rated]
Particle Size5 μm
pH2.5 to 7.5
Pore Size300 Å
Solvents100% Organic Solvents
Temperature30°C
USP TypeL59
Stationary PhaseDiol (OH)
Product LineMAbPac
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Column TypeSize Exclusion
PhaseSize Exclusion
MaterialPEEK
TechniqueBio-LC
Unit SizeEach
088790
also known as 88790
Full specifications
Each5 μm2.1 mm150 mmSize ExclusionRequest A Quote-
Column ChemistrySize Exclusion
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
Length (Metric)150 mm
Max. Pressure600 psi (40 bar)
Particle Size5 μm
pH2.5 to 7.5
Pore Size300 Å
Solvents100% Organic Solvents
Temperature30°C
USP TypeL59
Stationary PhaseProprietary Diol
Product LineMAbPac
Packing MaterialSpherical, High Purity, Porous Silica
Column TypeSize Exclusion
PhaseSize Exclusion
MaterialStainless Steel
TechniqueBio-LC
Unit SizeEach
088789Full specifications
Each5 μm2.1 mm300 mmSize ExclusionRequest A Quote-
Column ChemistrySize Exclusion
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
Length (Metric)300 mm
Max. Pressure1000 psi (70 bar)
Particle Size5 μm
pH2.5 to 7.5
Pore Size300 Å
Solvents100% Organic Solvents
Temperature30°C
USP TypeL59
Stationary PhaseProprietary Diol
Product LineMAbPac
Packing MaterialSpherical, High Purity, Porous Silica
Column TypeSize Exclusion
PhaseSize Exclusion
MaterialStainless Steel
Unit SizeEach
Showing 6 of 6
High-Performance SEC Columns for mAb Analysis
  • Hydrophilic bonded layer minimizes undesired interactions between the biomolecules and the stationary phase
  • Nonmetallic and biocompatible PEEK housing eliminates metal contamination from the column hardware
  • Stable surface bonding leads to low column bleed and compatibility with MS, ELSD and Thermo Scientific™ Dionex™ Corona™ CAD detection
  • Rugged column packing provides reliability, reproducibility and long column life
  • Columns are available in 5μm particle size and 150 and 300mm lengths, with an average pore diameter of 300Å
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text