Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Use Thermo Scientific™ MAbPac™ SEC-1 Size Exclusion Chromatography HPLC Columns for high-resolution separation of monoclonal antibody (mAb) analysis, including the monomers, aggregates and Fab and Fc fragments resulting from proteolysis. These size-exclusion chromatography (SEC) columns provide superior performance, even under nondenaturing conditions, and in high- and low-salt mobile phases and volatile eluents.
Column Chemistry | Proprietary Diol |
Column Format | Analytical Column |
Diameter (Metric) | 7.8 mm |
Flow Rate | 760 to 1000 μL/min |
For Use With | Protein Mol wt Range 10 to 1000 kDa |
Length (Metric) | 300 mm |
Max. Pressure | 1000 psi (70 bar) |
Particle Size | 5 μm |
pH | 2.5 to 7.5 |
Pore Size | 300 Å |
Solvents | 100% Organic Solvents |
Temperature | 30°C |
Stationary Phase | Diol (OH) |
Product Line | MAbPac |
Packing Material | Spherical, Fully Porous, Ultrapure Silica |
Column Type | Size Exclusion |
Phase | Size Exclusion |
Technique | Bio-LC |
Unit Size | Each |
Column Chemistry | Proprietary Diol |
Column Format | Analytical Column |
Diameter (Metric) | 4 mm |
Length (Metric) | 300 mm |
Max. Pressure | 1000 psi (70 bar) |
Particle Size | 5 μm |
pH | 2.5 to 7.5 |
Pore Size | 300 Å |
Solvents | 100% Organic Solvents |
Temperature | 30°C |
USP Type | L59 |
Stationary Phase | Diol (OH) |
Product Line | MAbPac |
Packing Material | Spherical, Fully Porous, Ultrapure Silica |
Column Type | Size Exclusion |
Phase | Size Exclusion |
Material | PEEK |
Technique | Bio-LC |
Unit Size | Each |
Column Chemistry | Proprietary Diol |
Column Format | Guard Column |
Diameter (Metric) | 4 mm |
Length (Metric) | 50 mm |
Max. Pressure | [Not Rated] |
Particle Size | 5 μm |
pH | 2.5 to 7.5 |
Pore Size | 300 Å |
Solvents | 100% Organic Solvents |
Temperature | 30°C |
USP Type | L59 |
Stationary Phase | Diol (OH) |
Product Line | MAbPac |
Packing Material | Spherical, Fully Porous, Ultrapure Silica |
Column Type | Size Exclusion |
Phase | Size Exclusion |
Material | PEEK |
Technique | Bio-LC |
Unit Size | Each |
Column Chemistry | Proprietary Diol |
Column Format | Analytical Column |
Diameter (Metric) | 4 mm |
Length (Metric) | 150 mm |
Max. Pressure | 600 psi (40 bar) |
Particle Size | 5 μm |
pH | 2.5 to 7.5 |
Pore Size | 300 Å |
Solvents | 100% Organic Solvents |
Temperature | 30°C |
USP Type | L59 |
Stationary Phase | Diol (OH) |
Product Line | MAbPac |
Packing Material | Spherical, Fully Porous, Ultrapure Silica |
Column Type | Size Exclusion |
Phase | Size Exclusion |
Material | PEEK |
Technique | Bio-LC |
Unit Size | Each |
Column Chemistry | Size Exclusion |
Column Format | Analytical Column |
Diameter (Metric) | 2.1 mm |
Length (Metric) | 150 mm |
Max. Pressure | 600 psi (40 bar) |
Particle Size | 5 μm |
pH | 2.5 to 7.5 |
Pore Size | 300 Å |
Solvents | 100% Organic Solvents |
Temperature | 30°C |
USP Type | L59 |
Stationary Phase | Proprietary Diol |
Product Line | MAbPac |
Packing Material | Spherical, High Purity, Porous Silica |
Column Type | Size Exclusion |
Phase | Size Exclusion |
Material | Stainless Steel |
Technique | Bio-LC |
Unit Size | Each |
Column Chemistry | Size Exclusion |
Column Format | Analytical Column |
Diameter (Metric) | 2.1 mm |
Length (Metric) | 300 mm |
Max. Pressure | 1000 psi (70 bar) |
Particle Size | 5 μm |
pH | 2.5 to 7.5 |
Pore Size | 300 Å |
Solvents | 100% Organic Solvents |
Temperature | 30°C |
USP Type | L59 |
Stationary Phase | Proprietary Diol |
Product Line | MAbPac |
Packing Material | Spherical, High Purity, Porous Silica |
Column Type | Size Exclusion |
Phase | Size Exclusion |
Material | Stainless Steel |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Particle Size | Diameter (Metric) | Length (Metric) | Column Type | Price (USD) | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
088460 | Each | 5 μm | 7.8 mm | 300 mm | Size Exclusion | 2,561.27 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
074696 | Each | 5 μm | 4 mm | 300 mm | Size Exclusion | 2,013.09 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
074697 | Each | 5 μm | 4 mm | 50 mm | Size Exclusion | 670.64 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
075592 | Each | 5 μm | 4 mm | 150 mm | Size Exclusion | 1,526.73 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
088790 also known as 88790 | Each | 5 μm | 2.1 mm | 150 mm | Size Exclusion | Request A Quote | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
088789 | Each | 5 μm | 2.1 mm | 300 mm | Size Exclusion | Request A Quote | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|