Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Obtain fast and highly robust, reproducible HPLC gradients using Thermo Scientific™ CX-1 pH Gradient Buffers. Simple to optimize and easily automated, these buffers are applicable for HPLC separation of the majority of monoclonal antibody (mAb) biotherapeutics drugs. CX-1 pH Gradient Buffers are ready to use with existing HPLC columns without the need to formulate mobile phases. The CX-1 buffers save time in method development, facilitate method transfer to QA/QC for a wide range of mAb charge variants through a generic HPLC-based approach to mAb characterization, and do not require time-consuming mobile phase adjustments.
Description | CX-1 pH Gradient Buffer B |
For Use With (Equipment) | LC Columns and Systems |
Product Line | Thermo Scientific |
Type | CX-1 pH Gradient Buffer B |
Volume (Metric) | 125 mL |
pH | 10.2 |
Quantity | 125 mL |
Unit Size | Each |
Description | CX-1 pH Gradient Buffer A |
For Use With (Equipment) | LC Columns and Systems |
Product Line | Thermo Scientific |
Type | CX-1 pH Gradient Buffer A |
Volume (Metric) | 125 mL |
pH | 5.6 |
Quantity | 125 mL |
Unit Size | Each |
Description | CX-1 pH Gradient Buffer A |
For Use With (Equipment) | LC Columns and Systems |
Product Line | Thermo Scientific |
Type | CX-1 pH Gradient Buffer A |
Volume (Metric) | 250 mL |
pH | 5.6 |
Quantity | 250 mL |
Unit Size | Each |
Description | CX-1 pH Gradient Buffer B |
For Use With (Equipment) | LC Columns and Systems |
Product Line | Thermo Scientific |
Type | CX-1 pH Gradient Buffer B |
Volume (Metric) | 250 mL |
pH | 10.2 |
Quantity | 250 mL |
Unit Size | Each |
Description | CX-1 pH Gradient Buffer A |
For Use With (Equipment) | LC Columns and Systems |
Product Line | Thermo Scientific |
Type | CX-1 pH Gradient Buffer A |
pH | 5.6 |
Quantity | 500 mL |
Unit Size | Each |
Description | CX-1 pH Gradient Buffer B |
For Use With (Equipment) | LC Columns and Systems |
Product Line | Thermo Scientific |
Type | CX-1 pH Gradient Buffer B |
pH | 10.2 |
Quantity | 500 mL |
Unit Size | Each |
Description | CX-1 pH Gradient Buffer A |
For Use With (Equipment) | HPLC Columns Including ProPac Elite WCX and MAbPac SCX HPLC Columns |
Product Line | Thermo Scientific |
Type | CX-1 pH Gradient Buffer A |
Volume (Metric) | 1000 mL |
pH | 5.6 |
Quantity | 1 L |
Unit Size | Each |
Description | CX-1 pH Gradient Buffer B |
For Use With (Equipment) | HPLC Columns Including ProPac Elite WCX and MAbPac SCX HPLC Columns |
Product Line | Thermo Scientific |
Type | CX-1 pH Gradient Buffer B |
Volume (Metric) | 1000 mL |
pH | 10.2 |
Quantity | 1 L |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | pH | Quantity | Price (USD) | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
083275 | Each | CX-1 pH Gradient Buffer B | 10.2 | 125 mL | 185.63 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
083273 | Each | CX-1 pH Gradient Buffer A | 5.6 | 125 mL | 185.63 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
085346 | Each | CX-1 pH Gradient Buffer A | 5.6 | 250 mL | 297.45 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
085348 | Each | CX-1 pH Gradient Buffer B | 10.2 | 250 mL | 297.45 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
302779 | Each | CX-1 pH Gradient Buffer A | 5.6 | 500 mL | 528.93 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
302780 | Each | CX-1 pH Gradient Buffer B | 10.2 | 500 mL | 505.45 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
303274 | Each | CX-1 pH Gradient Buffer A | 5.6 | 1 L | 820.00 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
303275 | Each | CX-1 pH Gradient Buffer B | 10.2 | 1 L | 916.97 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
pH buffer solutions can be used to generate highly reproducible, linear pH gradients in cation exchange chromatography.
Unlike traditional cation exchange chromatography using salt gradients, it is possible to predict the pH and the expected retention of the charge variants and use a narrow pH range to get a higher resolution separation.
Thermo Scientific pH buffer concentrates used in the pH gradient platform can be purchased individually or as a pair, in quantities of 125mL or 250mL. For added convenience, the 125mL buffers can also be bundled with columns in a number of specifically preconfigured kits.