Informational:Informational:Save up to 31% on your order of cell culture products. View promotion details »
Gibco™

PBS, pH 7.4

Catalog number: 10010072
Have Questions?
Gibco™

PBS, pH 7.4

Catalog number: 10010072
Have Questions?
Catalog Number
10010072
Unit Size
6 x 1,000 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1,000 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
500 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
10 x 500 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
100100726 x 1,000 mL
-
100100311,000 mL
-
10010023500 mL
-
1001004910 x 500 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

PBS (phosphate buffered saline) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. PBS is formulated without calcium and magnesium for rinsing chelators from the culture before cell dissociation. We offer a variety of PBS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.


This PBS is manufactured as follows:
Without
• Calcium
• Magnesium
• Phenol Red


The complete formulation is available.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Specifications

Additive
No Sodium Pyruvate
Classification
Animal Origin-Free
Color
Clear
Dilution
1 X
Product Line
Gibco™
Product Type
Phosphate Buffered Saline (PBS)
Quantity
6 x 1000 mL
Solution Type
Phosphate Buffered Saline
For Use With (Application)
Washing Cells Before Dissociation; Diluting Cells for Counting and Preparing Reagents; Transporting Cells or Tissue
Green Features
Sustainable packaging
Osmolality
280 - 315 mOsm/kg
pH
7.4
Phenol Red Indicator
No Phenol Red
Shipping Condition
Room Temperature
Sterility
Sterile-filtered
Form
Liquid
Manufacturing Quality
cGMP-compliant under the ISO 13485 standard

Contents & Storage

Storage conditions: 15°C to 30°C
Shipping conditions: Room temperature
Shelf life: 24 months from date of manufacture

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text