AcroMetrix™ Solid Tumor Controls (RUO)

Catalog number: 10024432
Have Questions?

AcroMetrix™ Solid Tumor Controls (RUO)

Catalog number: 10024432
Have Questions?
Validate and assess run-to-run solid tumor assay variation. Intended for use with next generation sequencing (NGS), qualitative and quantitative PCR research assays that are designed to identify fusions, CNVs, SNVs and indels. For research use only.
 
Catalog Number
10024432
Unit Size
Each
Description
AcroMetrix Core Fusion RNA Control (RUO)
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionAcroMetrix Core Fusion RNA Control (RUO)
PackagingOne kit contains 8 vials per kit x 8 uL per vial
Unit SizeEach
Catalog Number
Unit Size
Each
Description
AcroMetrix Core Hotspot-CNV DNA Control (RUO)
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionAcroMetrix Core Hotspot-CNV DNA Control (RUO)
PackagingOne kit contains 8 vials per kit x 8 uL per vial
Unit SizeEach
Catalog Number
Unit Size
Each
Description
AcroMetrix Hotspot Frequency Ladder (RUO)
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionAcroMetrix Hotspot Frequency Ladder (RUO)
PackagingOne kit contains 7 vials per kit x 30 uL per vial
Unit SizeEach
Showing 3 of 3
Catalog NumberSpecificationsUnit SizeDescriptionPrice (USD)AvailabilityQuantity
10024432Full specifications
EachAcroMetrix Core Fusion RNA Control (RUO)
-
DescriptionAcroMetrix Core Fusion RNA Control (RUO)
PackagingOne kit contains 8 vials per kit x 8 uL per vial
Unit SizeEach
10024431Full specifications
EachAcroMetrix Core Hotspot-CNV DNA Control (RUO)
-
DescriptionAcroMetrix Core Hotspot-CNV DNA Control (RUO)
PackagingOne kit contains 8 vials per kit x 8 uL per vial
Unit SizeEach
10026229Full specifications
EachAcroMetrix Hotspot Frequency Ladder (RUO)
-
DescriptionAcroMetrix Hotspot Frequency Ladder (RUO)
PackagingOne kit contains 7 vials per kit x 30 uL per vial
Unit SizeEach
Showing 3 of 3

AcroMetrix Core Fusion RNA Control (RUO)

  • Includes SLC34A2-ROS1 & EML4-ALK fusions


AcroMetrix Core Hotspot-CNV DNA Control (RUO)

  • Five hotspots and 1 CNV representative across various cancers


AcroMetrix Hotspot Frequency Ladder (RUO)

  • 555 hotspots include 504 SNV, 2 MNV, 29 DEL, 19 INS and 1 complex indel all in one control at 50%, 25%, 15%, 10%, 5%, 2.5% variant allele frequency
  • Includes a negative control at 0% variant allele frequency
FAQs
For Research Use Only. Not for use in diagnostic procedures.

Contents & Storage

-70°C

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text