Anthracene, 99%
Anthracene, 99%
Anthracene, 99%
Thermo Scientific Chemicals

Anthracene, 99%

CAS: 120-12-7 | C14H10 | 178.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
View all
Glass Bottle
Catalog number 104860050
Price (USD)
33.65
Special offer
Online exclusive
39.40
Save 5.75 (15%)
Each
-
Add to cart
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
33.65
Special offer
Online exclusive
39.40
Save 5.75 (15%)
Each
Add to cart
Chemical Identifiers
CAS120-12-7
IUPAC Nameanthracene
Molecular FormulaC14H10
InChI KeyMWPLVEDNUUSJAV-UHFFFAOYSA-N
SMILESC1=CC2=CC3=CC=CC=C3C=C2C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=98.5 %
Appearance (Form)Powder or crystals
Appearance (Color)White to yellow to beige to green-beige
Melting point214°C to 219°C
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0446055Certificate of AnalysisMay 17, 2025104860100, 104860025, 104861000, 104865000, 104860050
A0432680Certificate of AnalysisJan 29, 2024104860100, 104860025, 104861000, 104865000, 104860050
A0457226Certificate of AnalysisOct 25, 2023104860100, 104860025, 104861000, 104865000, 104860050, 10486025
2444902Certificate of AnalysisMay 26, 202310486025, 104865000, 104861000, 104860050, 104860100, 104860025
2193120Certificate of AnalysisMay 26, 202310486025, 104865000, 104861000, 104860050, 104860100, 104860025
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection