4-Bromobenzaldehyde, 99%
4-Bromobenzaldehyde, 99%
4-Bromobenzaldehyde, 99%
4-Bromobenzaldehyde, 99%
Thermo Scientific Chemicals

4-Bromobenzaldehyde, 99%

CAS: 1122-91-4 | C7H5BrO | 185.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
25 g
Packaging:
Glass bottle
Catalog number 106675000
Price (EUR)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1122-91-4
IUPAC Name4-bromobenzaldehyde
Molecular FormulaC7H5BrO
InChI KeyZRYZBQLXDKPBDU-UHFFFAOYSA-N
SMILESBrC1=CC=C(C=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline powder or crystals and/or
Melting point55°C to 59°C
Appearance (Color)White to light beige
Appearance (Form)chunks or fused solid
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0472440Certificate of AnalysisMay 17, 2025106670250, 106671000, 106675000
A0456775Certificate of AnalysisOct 11, 2023106670250, 106671000, 106675000
A0426660Certificate of AnalysisAug 25, 2023106670250, 106671000, 106675000
A0395094Certificate of AnalysisDec 15, 2022106670250, 106671000, 106675000
A0405819Certificate of AnalysisDec 12, 2022106670250, 106671000, 106675000
5 results displayed, search above for a specific certificate

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Skin Corrosion/Irritation
Category 2
Serious Eye Damage/Eye Irritation
Category 2
Specific target organ toxicity - (single exposure)
Category 3
Label Elements
Signal Word

Warning

Hazard Statements

H315 - Causes skin irritation

H319 - Causes serious eye irritation

H335 - May cause respiratory irritation

Precautionary Statements

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

Share catalog number, name or link

1x1 image pixel for data collection