n-Butyl acetate, 99+%, extra pure
n-Butyl acetate, 99+%, extra pure
n-Butyl acetate, 99+%, extra pure
n-Butyl acetate, 99+%, extra pure
Thermo Scientific Chemicals

n-Butyl acetate, 99+%, extra pure

CAS: 123-86-4 | C6H12O2 | 116.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
25 L
Packaging:
View all
Glass Bottle
Catalog number 107750025
Price (USD)
107.65
Special offer
Online exclusive
Ends: 26-Sep-2025
127.00
Save 19.35 (15%)
Each
-
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
107.65
Special offer
Online exclusive
Ends: 26-Sep-2025
127.00
Save 19.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS123-86-4
IUPAC Namebutyl acetate
Molecular FormulaC6H12O2
InChI KeyDKPFZGUDAPQIHT-UHFFFAOYSA-N
SMILESCCCCOC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Liquid
Water=<0.1 % (K.F.)
Refractive index1.3930 to 1.3950 (20°C, 589 nm)
Appearance (Color)Clear colorless
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Los clientes que vieron este artículo también vieron



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
2534896Certificate of AnalysisMay 24, 2025107750010, 107750250, 10227023, 107750025
2523716Certificate of AnalysisJan 22, 2025107750010, 107750250, 107750025
2412043Certificate of AnalysisDec 23, 2024107750010, 107750250, 107750025
2418681Certificate of AnalysisDec 23, 2024107750010, 107750250, 107750025
2418680Certificate of AnalysisDec 23, 2024107750010, 107750250, 107750025
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection