4-terc-Butilfenol, 97 %, Thermo Scientific Chemicals
4-terc-Butilfenol, 97 %, Thermo Scientific Chemicals
4-terc-Butilfenol, 97 %, Thermo Scientific Chemicals
4-terc-Butilfenol, 97 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-terc-Butilfenol, 97 %, Thermo Scientific Chemicals

CAS: 98-54-4 | C10H14O | 150.22 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
5 g
1 kg
2.5 kg
Número de catálogo 108000050
Precio (EUR)
17,68
Precio exclusivo en nuestra web
20,80
Ahorro 3,12 (15%)
Each
-
Añadir al carro de la compra
Cantidad:
5 g
Recurring order eligible. Learn more »
Pedido a granel o personalizado
Precio (EUR)
17,68
Precio exclusivo en nuestra web
20,80
Ahorro 3,12 (15%)
Each
Añadir al carro de la compra
Identificadores químicos
CAS98-54-4
IUPAC Name4-tert-butylphenol
Molecular FormulaC10H14O
InChI KeyQHPQWRBYOIRBIT-UHFFFAOYSA-N
SMILESCC(C)(C)C1=CC=C(O)C=C1
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)White to light beige
Infrared spectrumConforms
GC>=96.0 %
Water=<0.5 % (K.F.)
Appearance (Form)Flakes or pastilles
Ver más
Ask our AI about this Product

Este producto de marca Thermo Scientific Chemicals formó parte originalmente de la cartera de productos Acros Organics. Alguna información sobre documentos y etiquetas puede referirse a la marca original. El producto original Acros Organics/código de artículo o referencia SKU no ha cambiado como parte de la transición de marca a Thermo Scientific Chemicals.

RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documentos y descargas

Certificados

N.º de loteCertificate TypeDateCatalog Number(s)
A0432201Certificate of Analysis08 ene 2024108000050, 108000010, 108000025
A0430439Certificate of Analysis27 nov 2023108000050, 108000010, 108000025
A0424914Certificate of Analysis25 ago 2023108000050, 108000010, 108000025
A0395013Certificate of Analysis14 sept 2020108000050, 108000010, 108000025
A0421936Certificate of Analysis26 ago 2020108000010, 108000025, 108000050
Se muestran 5 resultados, busque arriba un certificado específico

Hojas de datos de seguridad

Seguridad y manipulación

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Skin Corrosion/Irritation
Category 2
Serious Eye Damage/Eye Irritation
Category 1
Reproductive Toxicity
Category 2
Chronic aquatic toxicity
Category 1
Label Elements
Signal Word

Danger

Hazard Statements

H315 - Causes skin irritation

H318 - Causes serious eye damage

H361f - Suspected of damaging fertility

H410 - Very toxic to aquatic life with long lasting effects

Precautionary Statements

P201 - Obtain special instructions before use

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

P310 - Immediately call a POISON CENTER or doctor/physician

Compartir número de catálogo, nombre o enlace.

1x1 image pixel for data collection