Citric acid, 99%, pure, anhydrous
Citric acid, 99%, pure, anhydrous
Citric acid, 99%, pure, anhydrous
Citric acid, 99%, pure, anhydrous
Thermo Scientific Chemicals

Citric acid, 99%, pure, anhydrous

CAS: 77-92-9 | C6H8O7 | 192.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 kg
1 kg
5 kg
Packaging:
View all
Plastic Bottle
Catalog number 110450010
Price (USD)
83.65
Online Exclusive
92.70
Save 9.05 (10%)
Each
-
Add to cart
Quantity:
1 kg
Packaging:
Plastic Bottle
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
83.65
Online Exclusive
92.70
Save 9.05 (10%)
Each
Add to cart
Chemical Identifiers
CAS77-92-9
IUPAC Name2-hydroxypropane-1,2,3-tricarboxylic acid
Molecular FormulaC6H8O7
InChI KeyKRKNYBCHXYNGOX-UHFFFAOYSA-N
SMILESOC(=O)CC(O)(CC(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals
Titration with NaOH>=98.5 %
Infrared spectrumConforms
Appearance (Color)White
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Will the Cells-to-CT system work with my special cell line?
Can Single Cell-to-CT kit reagents be subjected to freeze–thaw cycles?
How do I remove gDNA from my Cells-to-CT reaction?
Can cells sorted by flow cytometry be used with the Single Cell-to-CT qRT-PCR Kit (Cat. No. 4458237)?
Which cell lines have been tested to work with the Cells-to-CT system?
+Show more FAQs for this product
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0427603Certificate of AnalysisAug 28, 2023110450250, 110450050, 110450010
A0453307Certificate of AnalysisAug 25, 2023110450250, 110450050, 110450010
A0393282Certificate of AnalysisDec 15, 2022110450250, 110450050, 110450010
A0393283Certificate of AnalysisDec 15, 2022110450250, 110450050, 110450010
A0410335Certificate of AnalysisDec 13, 2022110450250, 110450050, 110450010
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link