o-Cresol, 99%, Thermo Scientific Chemicals
o-Cresol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

o-Cresol, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
100 g
500 g
2.5 kg
10 kg
Packaging:
View all
Glass bottle
Catalog number 110550010
Price (USD)/ Each
144.65
Online exclusive
160.00 
Save 15.35 (10%)
-
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
144.65
Online exclusive
160.00 
Save 15.35 (10%)
Add to cart
o-Cresol, 99%, Thermo Scientific Chemicals
Catalog number110550010
Price (USD)/ Each
144.65
Online exclusive
160.00 
Save 15.35 (10%)
-
Add to cart
Chemical Identifiers
CAS95-48-7
IUPAC Name2-methylphenol
Molecular FormulaC7H8O
InChI KeyQWVGKYWNOKOFNN-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
AppearanceMay darken on storage
Appearance (Form)Low melting solid or liquid
GC>=98.5 %
Appearance (Color)Colorless to amber
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only