Search Thermo Fisher Scientific
Solve more laboratory challenges, improve workflow, and minimize downtime when performing carbon, hydrogen, nitrogen, sulfur and oxygen (CHNS/O) analyses with the Thermo Scientific™ FlashSmart™ Elemental Analyzers. Based on the modified Dumas Method, the FlashSmart Elemental Analyzer is highly flexible, offering modularity that enables over 20 configurations in one instrument. Besides expanding your analytical capabilities, this analyzer features powerful software that supports automation and precise reports, making the system easier to use from start to finish.
Type | Elemental Analyzer |
Detector Type | Thermal Conductivity Detector |
For Use With (Application) | Organic Chemistry, Pharmaceuticals, Petro-Chemistry, Material Characterization, Energy |
Description | FlashSmart CHNS |
Unit Size | Each |
Type | Elemental Analyzer |
Detector Type | Thermal Conductivity Detector |
For Use With (Application) | Food Safety |
Description | FlashSmart N/PROTEIN |
Unit Size | Each |
Type | Elemental Analyzer |
Detector Type | Thermal Conductivity Detector |
For Use With (Application) | Agronomy, Environmental Science, Marine Science, Material Characterization |
Description | FlashSmart NC SOIL |
Unit Size | - |
Type | Elemental Analyzer |
Detector Type | Thermal Conductivity Detector |
For Use With (Application) | Organic Chemistry, Pharmaceuticals |
Description | FlashSmart CHN |
Unit Size | Each |
Type | Elemental Analyzer |
Detector Type | Thermal Conductivity Detector |
For Use With (Application) | Organic Chemistry, Pharmaceuticals, Environmental Science |
Description | FlashSmart CHN/O |
Unit Size | Each |
Type | Elemental Analyzer |
Detector Type | Thermal Conductivity Detector |
For Use With (Application) | Organic Chemistry, Pharmaceuticals, Petro-Chemistry, Material Characterization, Energy |
Description | FlashSmart CHNS/O |
Unit Size | Each |
Type | Elemental Analyzer |
Detector Type | Thermal Conductivity Detector |
For Use With (Application) | Agronomy, Marine Science, Material Characterization |
Description | FlashSmart N ORG. |
Unit Size | Each |
Type | Elemental Analyzer |
Detector Type | Thermal Conductivity Detector |
For Use With (Application) | Agronomy, Marine Science, Material Characterization |
Description | FlashSmart NC ORG. |
Unit Size | Each |
Type | Elemental Analyzer |
Detector Type | Thermal Conductivity Detector |
For Use With (Application) | Environmental Science, Agronomy, Marine Science, Material Characterization |
Description | FlashSmart NCS |
Unit Size | Each |
Type | Elemental Analyzer |
Detector Type | Thermal Conductivity Detector |
For Use With (Application) | Petro-Chemistry, Energy |
Description | FlashSmart N LUBRICANT |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | For Use With (Application) | Price (USD) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
11206115 | Each | FlashSmart CHNS | Organic Chemistry | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
11206100 | Each | FlashSmart N/PROTEIN | Food Safety | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
11206105 | - | FlashSmart NC SOIL | Agronomy | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
11206110 | Each | FlashSmart CHN | Organic Chemistry | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
11206120 | Each | FlashSmart CHN/O | Organic Chemistry | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
11206125 | Each | FlashSmart CHNS/O | Organic Chemistry | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
11206130 | Each | FlashSmart N ORG. | Agronomy | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
11206135 | Each | FlashSmart NC ORG. | Agronomy | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
11206140 | Each | FlashSmart NCS | Environmental Science | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
11206145 | Each | FlashSmart N LUBRICANT | Petro-Chemistry | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Add automation to your daily organic elemental analysis for a wide range of applications without compromising precision and accuracy.