Thermo Scientific™
FlashSmart™ Elemental Analyzer
Catalog number: 11206135
Have Questions?
Thermo Scientific™
FlashSmart™ Elemental Analyzer
Catalog number: 11206135
Have Questions?
Solve more laboratory challenges, improve workflow, and minimize downtime when performing carbon, hydrogen, nitrogen, sulfur and oxygen (CHNS/O) analyses with the Thermo Scientific™ FlashSmart™ Elemental Analyzers. Based on the modified Dumas Method, the FlashSmart Elemental Analyzer is highly flexible, offering modularity that enables over 20 configurations in one instrument. Besides expanding your analytical capabilities, this analyzer features powerful software that supports automation and precise reports, making the system easier to use from start to finish.
Catalog Number
11206135
Unit Size
Each
Description
FlashSmart NC ORG.
For Use With (Application)
Agronomy
Price (USD)
Full specifications
TypeElemental Analyzer
Detector TypeThermal Conductivity Detector
For Use With (Application)Agronomy, Marine Science, Material Characterization
DescriptionFlashSmart NC ORG.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
FlashSmart N/PROTEIN
For Use With (Application)
Food Safety
Price (USD)
Full specifications
TypeElemental Analyzer
Detector TypeThermal Conductivity Detector
For Use With (Application)Food Safety
DescriptionFlashSmart N/PROTEIN
Unit SizeEach
Catalog Number
Unit Size
-
Description
FlashSmart NC SOIL
For Use With (Application)
Agronomy
Price (USD)
Full specifications
TypeElemental Analyzer
Detector TypeThermal Conductivity Detector
For Use With (Application)Agronomy, Environmental Science, Marine Science, Material Characterization
DescriptionFlashSmart NC SOIL
Unit Size-
Catalog Number
Unit Size
Each
Description
FlashSmart CHN
For Use With (Application)
Organic Chemistry
Price (USD)
Full specifications
TypeElemental Analyzer
Detector TypeThermal Conductivity Detector
For Use With (Application)Organic Chemistry, Pharmaceuticals
DescriptionFlashSmart CHN
Unit SizeEach
Catalog Number
Unit Size
Each
Description
FlashSmart CHNS
For Use With (Application)
Organic Chemistry
Price (USD)
Full specifications
TypeElemental Analyzer
Detector TypeThermal Conductivity Detector
For Use With (Application)Organic Chemistry, Pharmaceuticals, Petro-Chemistry, Material Characterization, Energy
DescriptionFlashSmart CHNS
Unit SizeEach
Catalog Number
Unit Size
Each
Description
FlashSmart CHN/O
For Use With (Application)
Organic Chemistry
Price (USD)
Full specifications
TypeElemental Analyzer
Detector TypeThermal Conductivity Detector
For Use With (Application)Organic Chemistry, Pharmaceuticals, Environmental Science
DescriptionFlashSmart CHN/O
Unit SizeEach
Catalog Number
Unit Size
Each
Description
FlashSmart CHNS/O
For Use With (Application)
Organic Chemistry
Price (USD)
Full specifications
TypeElemental Analyzer
Detector TypeThermal Conductivity Detector
For Use With (Application)Organic Chemistry, Pharmaceuticals, Petro-Chemistry, Material Characterization, Energy
DescriptionFlashSmart CHNS/O
Unit SizeEach
Catalog Number
Unit Size
Each
Description
FlashSmart N ORG.
For Use With (Application)
Agronomy
Price (USD)
Full specifications
TypeElemental Analyzer
Detector TypeThermal Conductivity Detector
For Use With (Application)Agronomy, Marine Science, Material Characterization
DescriptionFlashSmart N ORG.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
FlashSmart NCS
For Use With (Application)
Environmental Science
Price (USD)
Full specifications
TypeElemental Analyzer
Detector TypeThermal Conductivity Detector
For Use With (Application)Environmental Science, Agronomy, Marine Science, Material Characterization
DescriptionFlashSmart NCS
Unit SizeEach
Catalog Number
Unit Size
Each
Description
FlashSmart N LUBRICANT
For Use With (Application)
Petro-Chemistry
Price (USD)
Full specifications
TypeElemental Analyzer
Detector TypeThermal Conductivity Detector
For Use With (Application)Petro-Chemistry, Energy
DescriptionFlashSmart N LUBRICANT
Unit SizeEach
Showing 10 of 14Show more
Catalog NumberSpecificationsUnit SizeDescriptionFor Use With (Application)Price (USD)
11206135Full specifications
EachFlashSmart NC ORG.AgronomyRequest A Quote
TypeElemental Analyzer
Detector TypeThermal Conductivity Detector
For Use With (Application)Agronomy, Marine Science, Material Characterization
DescriptionFlashSmart NC ORG.
Unit SizeEach
11206100Full specifications
EachFlashSmart N/PROTEINFood SafetyRequest A Quote
TypeElemental Analyzer
Detector TypeThermal Conductivity Detector
For Use With (Application)Food Safety
DescriptionFlashSmart N/PROTEIN
Unit SizeEach
11206105Full specifications
-FlashSmart NC SOILAgronomyRequest A Quote
TypeElemental Analyzer
Detector TypeThermal Conductivity Detector
For Use With (Application)Agronomy, Environmental Science, Marine Science, Material Characterization
DescriptionFlashSmart NC SOIL
Unit Size-
11206110Full specifications
EachFlashSmart CHNOrganic ChemistryRequest A Quote
TypeElemental Analyzer
Detector TypeThermal Conductivity Detector
For Use With (Application)Organic Chemistry, Pharmaceuticals
DescriptionFlashSmart CHN
Unit SizeEach
11206115Full specifications
EachFlashSmart CHNSOrganic ChemistryRequest A Quote
TypeElemental Analyzer
Detector TypeThermal Conductivity Detector
For Use With (Application)Organic Chemistry, Pharmaceuticals, Petro-Chemistry, Material Characterization, Energy
DescriptionFlashSmart CHNS
Unit SizeEach
11206120Full specifications
EachFlashSmart CHN/OOrganic ChemistryRequest A Quote
TypeElemental Analyzer
Detector TypeThermal Conductivity Detector
For Use With (Application)Organic Chemistry, Pharmaceuticals, Environmental Science
DescriptionFlashSmart CHN/O
Unit SizeEach
11206125Full specifications
EachFlashSmart CHNS/OOrganic ChemistryRequest A Quote
TypeElemental Analyzer
Detector TypeThermal Conductivity Detector
For Use With (Application)Organic Chemistry, Pharmaceuticals, Petro-Chemistry, Material Characterization, Energy
DescriptionFlashSmart CHNS/O
Unit SizeEach
11206130Full specifications
EachFlashSmart N ORG.AgronomyRequest A Quote
TypeElemental Analyzer
Detector TypeThermal Conductivity Detector
For Use With (Application)Agronomy, Marine Science, Material Characterization
DescriptionFlashSmart N ORG.
Unit SizeEach
11206140Full specifications
EachFlashSmart NCSEnvironmental ScienceRequest A Quote
TypeElemental Analyzer
Detector TypeThermal Conductivity Detector
For Use With (Application)Environmental Science, Agronomy, Marine Science, Material Characterization
DescriptionFlashSmart NCS
Unit SizeEach
11206145Full specifications
EachFlashSmart N LUBRICANTPetro-ChemistryRequest A Quote
TypeElemental Analyzer
Detector TypeThermal Conductivity Detector
For Use With (Application)Petro-Chemistry, Energy
DescriptionFlashSmart N LUBRICANT
Unit SizeEach
Showing 10 of 14Show more
FlashSmart Elemental Analyzer Features
Add automation to your daily organic elemental analysis for a wide range of applications without compromising precision and accuracy.
  • Simple design and usability
  • Improved CHNS/O workflow analysis
  • Higher sample throughput
  • Higher modularity
  • Dedicated features