Dynabeads™ Untouched™ Mouse CD4 Cells Kit
Dynabeads™ Untouched™ Mouse CD4 Cells Kit
Invitrogen™

Dynabeads™ Untouched™ Mouse CD4 Cells Kit

Dynabeads™ Untouched™ Mouse CD4 Cells contains both magnetic beads and an Antibody Mix to deplete all leucocytes from mouse spleenRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
11416D4 mL
11415D2 x 10 mL
Catalog number 11416D
Price (USD)
325.65
Online Exclusive
350.00
Save 24.35 (7%)
Each
In stock
Add to cart
Quantity:
4 mL
Recurring order eligible. Learn more »
Price (USD)
325.65
Online Exclusive
350.00
Save 24.35 (7%)
Each
Add to cart
Ask our AI about this Product

Dynabeads™ Untouched™ Mouse CD4 Cells contains both magnetic beads and an Antibody Mix to deplete all leucocytes from mouse spleen or lymph node cells, leaving only the target mouse CD4+ cells. The negatively isolated, untouched mouse CD4+ cells are pure, viable and ready to be used in any downstream application.

• Rapid isolation of untouched mouse CD4+ T-cells – no columns required
• Pure and viable mouse CD4+ T-cells ready for any downstream application

Excellent Yield, Purity, and Viability of Recovered Cells
The Mouse Depletion Dynabeads™ suspension contains uniform, superparamagnetic beads (4.5 μm diameter) that enable easy isolation of cells from any single cell suspension. This reagent is therefore especially well-suited for enriching tissue digests such as mouse spleen and lymph node cells. The Mouse Depletion Dynabeads™ are coated with a secondary polyclonal antibody that binds rat IgGs. The Antibody Mix contains a cocktail of rat IgGs that binds mouse CD8+ T cells, B cells, NK cells, monocytes/macrophages, dendritic cells, erythrocytes and granulocytes. The Antibody Mix is first added to the sample in a tube to bind the unwanted non-CD4+ cells. The cells are then washed to remove excess antibodies, and Mouse Depletion Dynabeads™ are added. After a short incubation, the bead-bound cells are then separated from unbound cells in 1 – 2 minutes using a magnet. The purified target mouse CD4+ cells in the supernatant can be transferred to a new tube for use in any downstream application. This fast and gentle isolation method does not require the use of columns, and helps ensure high purity, recovery and viability of the untouched CD4+ cells. The protocol can easily be scaled up according to your sample size.

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeT Cells (CD4)
Final Product TypeCells
Isolation TechnologyNegative Isolation
No. of CellsProcesses 2 x 108 cells
Product LineDYNAL™, Dynabeads™
Purity or Quality GradeResearch Grade
Quantity4 mL
ReactivityMouse
Sample TypeLymph Node, MNC, Spleen
Shipping ConditionRoom Temperature
Target SpeciesMouse
Diameter (Metric)4.5 μm
Product TypeCell Isolation Kit
Unit SizeEach
Contents & Storage
The Kit Contains: 4 mL Mouse Depletion Dynabeads™ and 0.4 mL Antibody Mix with monoclonal antibodies towards CD8, CD11b, CD16/32, CD45R and Ter-119.
Store at 2°C to 8°C.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Frequently asked questions (FAQs)

My Dynabeads magnetic beads are not pelleting well with the magnet. Do you have any suggestions for me?

Please review the following possibilities for why your Dynabeads magnetic beads are not pelleting:

- The solution is too viscous.
- The beads have formed aggregates because of protein-protein interaction.

Try these suggestions: - Increase separation time (leave tub on magnet for 2-5 minutes)
- Add DNase I to the lysate (~0.01 mg/mL)
- Increase the Tween 20 concentration to ~0.05% of the binding and/or washing buffer.
- Add up to 20 mM beta-merecaptoethanol to the binding and/or wash buffers.

Find additional tips, troubleshooting help, and resources within our Dynabeads Nucleic Acid Purification Support Center.

I have a long double-stranded DNA fragment I would like to isolate. What product do you recommend?

For biotin-labeled DNA that is less than 1 kb, we recommend you use Dynabeads M270 Streptavidin (Cat. No. 65305) and MyOne C1 magnetic beads (Cat. No. 65001). We recommend our Dynabeads KilobaseBINDER Kit (Cat. No. 60101), which is designed to immobilize long (>1 kb) double-stranded DNA molecules. The KilobaseBINDER reagent consists of M-280 Streptavidin-coupled Dynabeads magnetic beads along with a patented immobilization activator in the binding solution to bind to long, biotinylated DNA molecules for isolation. Please see the following link (https://www.thermofisher.com/us/en/home/life-science/dna-rna-purification-analysis/napamisc/capture-of-biotinylated-targets/immobilisation-of-long-biotinylated-dna-fragments.html) for more information in regards to long biotinylated DNA fragment isolation.

Find additional tips, troubleshooting help, and resources within our Dynabeads Nucleic Acid Purification Support Center.

Can I use Dynabeads magnetic beads to isolate single-stranded DNA templates?

Yes, Dynabeads magnetic beads can be used to isolate single-stranded DNA. Streptavidin Dynabeads magnetic beads can be used to target biotinylated DNA fragments, followed by denaturation of the double-stranded DNA and removal of the non-biotinylated strand. The streptavidin-coupled Dynabeads magnetic beads will not inhibit any enzymatic activity. This enables further handling and manipulation of the bead-bound DNA directly on the solid phase. Please see the following link (https://www.thermofisher.com/us/en/home/life-science/dna-rna-purification-analysis/napamisc/capture-of-biotinylated-targets/preparing-single-stranded-dna-templates.html) for more information in regards to single-stranded DNA capture.

Find additional tips, troubleshooting help, and resources within our Dynabeads Nucleic Acid Purification Support Center.

What is the magnetic susceptibility for Dynabeads magnetic beads?

Magnetic susceptibility is a measure of how quickly the beads will migrate to the magnet. This will depend on the iron content and the character of the iron oxide. The magnetic susceptibility given for the Dynabeads magnetic beads is the mass susceptibility, given either as cgs units/g or m^3/kg (the latter being an SI unit). For ferri- and ferromagnetic substances, the magnetic mass susceptibility is dependent upon the magnetic field strength (H), as the magnetization of such substances is not a linear function of H but approaches a saturation value with increasing field. For that reason, the magnetic mass susceptibility of the Dynabeads magnetic beads is determined by a standardized procedure under fixed conditions. The magnetic mass susceptibility given in our catalog is thus the SI unit. Conversion from Gaussian (cgs, emu) units into SI units for magnetic mass susceptibility is achieved by multiplying the Gaussian factor (emu/g or cgs/g) by 4 pi x 10^-3. The resulting unit is also called the rationalized magnetic mass susceptibility, which should be distinguished from the (SI) dimensionless magnetic susceptibility unit. In general, magnetic mass susceptibility is a measure of the force (Fz) influencing an object positioned in a nonhomogenous magnetic field. The magnetic mass susceptibility of the Dynabeads magnetic beads is measured by weighing a sample, and then subjecting the sample to a magnetic field of known strength. The weight (F1) is then measured, and compared to the weight of the sample when the magnetic field is turned off (F0). The susceptibility is then calculated as K x 10^-3 = [(F1-F0) x m x 0.335 x 10^6], where K is the mass susceptibility of the sample of mass m. The susceptibility is then converted to SI units.

Find additional tips, troubleshooting help, and resources within our Dynabeads Nucleic Acid Purification Support Center.

How can I determine coupling efficiency of Dynabeads magnetic beads?

There are different methods to check binding of ligands to the beads, including optical density (OD) measurement, fluorescent labeling, and radioactive labeling.

For OD measurement, you would measure the OD of the ligand before immobilization to the beads and compare it with the ligand concentration that is left in the supernatant after coating. This gives a crude measurement of how much protein has bound to the beads.

Protocol:

1.Set spectrophotometer to the right wavelength. As a blank, use the Coupling Buffer.
2.Measure the absorbance of the Pre-Coupling Solution. A further dilution may be necessary to read the absorbance, depending upon the amount of ligand added.
3.Measure the absorbance of the Post-Coupling Solution. A dilution may be necessary to read the absorbance.
4.Calculate the coupling efficiency, expressed as the % protein uptake, as follows. [(Pre-Coupling Solution x D) - (Post-Coupling Solution x D)] x 100/(Pre-Coupling Solution x D) where D = dilution factor.

For fluorescent labeling, we suggest negatively quantifying the amount of ligand bound by measuring ligand remaining in the coupling supernatant (compared to the original sample), rather than directly measuring the ligands on the beads. Add labeled ligand to the beads, and measure how much ligand is left in the supernatant (not bound to the beads). By comparing this with the total amount added in the first place, you can then calculate how much of the ligand that has been bound to the beads. Keep in mind that the Dynabeads magnetic beads are also autofluorescent, which is why direct measuring of fluorescence of the bead-bound ligands is not recommended, but rather this indirect approach. The label could be, for example, FITC/PE. Some researchers perform a direct approach with success (using a flow cytometer).

Radioactive labeling is the most sensitive method of the three, but it is also the most difficult one. It involves radioactively labeling a portion of the ligand. We use radiolabeled I-125 in tracer amounts and mix it with "cold" ligands in a known ratio before coupling. The absolute quantities for the ligand on the beads should be obtained by measuring the beads in a scintillation (gamma) counter and comparing the cpm with a standard.

Protocol:

1.Take out an appropriate amount of beads and wash the beads in 1 mL of binding buffer.
2.Pipette out desired amount of human IgG in a separate tube.
3.Mix the human IgG with I-125-labeled human IgG (30,000 - 100,000 cpm).
4.Dilute the mixture of human IgG and I-125-labeled human IgG to 100 mL in binding buffer.
5.Incubate for 30 minutes at room temperature and measure the cpm in a scintillation counter.
6.Wash the beads (with coating) four times, and measure cpm again.
The % binding is calculated by using the equation : (cpm after washing/cpm before washing)x100%.

Find additional tips, troubleshooting help, and resources within our Dynabeads Nucleic Acid Purification Support Center.

Figures

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3278755Certificate of AnalysisJul 11, 202511416D
3277970Certificate of AnalysisJul 10, 202511415D
3272625Certificate of AnalysisJul 02, 202511416D
3272461Certificate of AnalysisJul 02, 202511415D
3271067Certificate of AnalysisJun 30, 202511416D
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection