Informational:Still using Lipofectamine 2000? Try our upgraded product Lipofectamine 3000 Reagent.
Invitrogen™

Lipofectamine™ 2000 Transfection Reagent

Catalog number: 11668500
Have Questions?
Invitrogen™

Lipofectamine™ 2000 Transfection Reagent

Catalog number: 11668500
Have Questions?
Catalog Number
11668500
Unit Size
15 ml
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1.5 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
0.75 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
0.3 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
1166850015 ml
-
116680191.5 mL
-
116680270.75 mL
-
116680300.3 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Lipofectamine™ 2000 Transfection Reagent is a versatile transfection reagent that has been shown to effectively transfect the widest variety of adherent and suspension cell lines. Researchers use Lipofectamine™ 2000 Reagent for siRNA- and shRNA-based gene knockdown experiments, as well as for gene expression studies.

With Lipofectamine™ 2000 Transfection Reagent, you'll get:

• Exceptional transfection efficiency in the broadest range of cell lines and the highest levels of recombinant protein expression (see table)
• Superior performance for co-transfection of siRNA and plasmid DNA
• Proven efficacy in the presence of serum—eliminates the need to change media following transfection
• Reliable performance for high-throughput applications
• The best choice for establishing stable cell lines

A high-performance transfection reagent for gene expression and gene silencing
Lipofectamine™ 2000 Transfection Reagent works effectively with all common cell lines as well as with many challenging ones, and can be used in media with or without serum. For gene silencing, Lipofectamine™ 2000 Transfection Reagent's high-efficiency transfections provide the high levels of gene knockdown needed to produce convincing results. Lipofectamine™ 2000 Transfection Reagent is the number one choice for co-transfection, given its effectiveness for transfecting both siRNA and plasmid DNA. Lipofectamine™ 2000 Transfection Reagent is easy to use—simply mix with nucleic acid and add to cell culture.

Ideal for high-throughput work
Simplicity and speed combined with high transfection efficiency make Lipofectamine™ 2000 Transfection Reagent ideal for transient protein expression or high-throughput RNAi transfections. Transfection conditions can be easily established for automated or robotic systems used in such applications.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Type
Established Cell Lines, Stem Cells, Primary Cells, Hard-to-Transfect Cells
Format
6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks
Sample Type
Plasmid DNA, Synthetic siRNA, RNAi Plasmids (shRNA, miR)
Transfection Technique
Lipid-based Transfection
For Use With (Application)
Transfection
Green Features
Sustainable packaging
High-throughput Compatibility
High-throughput Compatible
Product Line
Lipofectamine™
Product Type
Transfection Reagent
Quantity
15 mL
Serum Compatible
Yes
Shipping Condition
Approved for shipment at Room Temperature or on Wet Ice

Contents & Storage

Contains one bottle (15 mL) Lipofectamine™ 2000 Reagent. Store at 2–8°C. Do not freeze.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text