Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Increase walk-away time and assay efficiency with Thermo Scientific™ Nalgene™ Disposable Polypropylene Robotic Reservoirs. Featuring a standard microplate format with 300mL volume, the convoluted design minimizes dead volume and has baffles to reduce splashing. The flat bottom version is designed to meet a variety of assay needs in both manual and automated environments.
Certifications/Compliance | Meets the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) |
Color | Natural |
Capacity (Metric) | 300 mL |
Dimensions (L x W x H) | 3.4 x 5 x 1.75 in. (86 x 128 x 43 mm) |
Material | Polypropylene |
Sterility | Sterile |
Type | Reservoir |
Height (English) | 1.75 in. |
Height (Metric) | 43 mm |
Length (English) | 3.4 in. |
Length (Metric) | 86 mm |
No. per Case | 40 |
No. per Pack | 4 |
Width (English) | 5 in. |
Width (Metric) | 128 mm |
Plate Compatibility | Single, 6-Well Plates, 12-Well Plates, 96-Well Plates, 384-Well Plates, 1536-Well Plates |
Bottom Shape | Flat |
Unit Size | Case of 40 |
Certifications/Compliance | Meets the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) |
Color | Natural |
Capacity (Metric) | 300 mL |
Dimensions (L x W x H) | 3.4 x 5 x 1.75 in. (86 x 128 x 43 mm) |
Material | Polypropylene |
Sterility | Non-sterile |
Type | Reservoir |
Height (English) | 1.75 in. |
Height (Metric) | 43 mm |
Length (English) | 3.4 in. |
Length (Metric) | 86 mm |
No. per Case | 40 |
No. per Pack | 4 |
Width (English) | 5 in. |
Width (Metric) | 128 mm |
Plate Compatibility | Single, 6-Well Plates, 12-Well Plates, 96-Well Plates, 384-Well Plates, 1536-Well Plates |
Bottom Shape | Flat |
Unit Size | Case of 40 |
Certifications/Compliance | American National Standards (ANSI) (ANSI/SBS 1-2004) |
Color | Natural |
Capacity (Metric) | 300 mL |
Dimensions (L x W x H) | 3.4 x 5 x 1.75 in. (86 x 128 x 43 mm) |
Material | Polypropylene |
Sterility | Non-sterile |
Type | Reservoir |
Height (English) | 1.75 in. |
Height (Metric) | 43 mm |
Length (English) | 3.4 in. |
Length (Metric) | 86 mm |
No. per Case | 40 |
No. per Pack | 4 |
Width (English) | 5 in. |
Width (Metric) | 128 mm |
Plate Compatibility | 96-Well Plates |
Bottom Shape | Convoluted |
Unit Size | Case of 40 |
Certifications/Compliance | Meets the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) |
Color | Natural |
Capacity (Metric) | 300 mL |
Dimensions (L x W x H) | 3.4 x 5 x 1.75 in. (86 x 128 x 43 mm) |
Material | Polypropylene |
Sterility | Sterile |
Type | Reservoir |
Height (English) | 1.75 in. |
Height (Metric) | 43 mm |
Length (English) | 3.4 in. |
Length (Metric) | 86 mm |
No. per Case | 40 |
No. per Pack | 4 |
Width (English) | 5 in. |
Width (Metric) | 128 mm |
Plate Compatibility | 96-Well Plates |
Bottom Shape | Convoluted |
Unit Size | Case of 40 |
Catalog Number | Specifications | Unit Size | Bottom Shape | Sterility | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
1200-1301 | Case of 40 | Flat | Sterile | 350.65 Online price 356.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
1200-1300 | Case of 40 | Flat | Non-sterile | 307.65 Online price 312.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
1200-2300 | Case of 40 | Convoluted | Non-sterile | 314.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
1200-2301 | Case of 40 | Convoluted | Sterile | 349.65 Online price 356.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|