PureLink™ Plant RNA Reagent
Shipment of product is currently delayed.
Product Image
Invitrogen™

PureLink™ Plant RNA Reagent

Plant RNA Reagent is a single-component solution optimized for the isolation of high yields of high-quality total RNA from aRead more
Have Questions?
Catalog NumberQuantity
12322012100 mL
Catalog number 12322012
Price (USD)
-
Quantity:
100 mL
Ask our AI about this Product
Plant RNA Reagent is a single-component solution optimized for the isolation of high yields of high-quality total RNA from a variety of plant tissues, especially those rich in polyphenolics (e.g., conifer needles) or starch (e.g., potato tuber or seeds). Features of the Plant RNA Reagent:

• Isolate high-quality RNA from difficult plant materials
• Use for small-scale and large-scale RNA isolations

Improved quality and purity of plant RNA
Plant RNA Reagent is optimized to have less sample DNA contamination, to be twice as effective as TRIzol™ Reagent, and have a simplified RNA extraction protocol for minimized time and labor. The end result is high-yield, high-purity RNA (determined by A260/A280 and gel analysis) from a variety of plant material sources.

Works for milligram to gram quantities of starting material
Plant RNA Reagent provides greater flexibility for sample size and processing formats, and can be used for both small-scale (up to 0.1 g plant tissue) and large-scale (0.1–5 g plant tissue) total RNA isolation. The amount supplied (100 mL) is sufficient for 200 reactions using 100 mg of tissue, or 4 reactions using 5 g of tissue.

Isolated RNA is ideal for a variety of applications and products:

cDNA library construction kits
Real-Time PCR (qPCR)
Northern blotting kits & reagents
Poly(A) selection kit
RNase control reagents & assays
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Final Product TypeTotal RNA
For Use With (Application)Reverse Transcriptase PCR (RT-PCR), cDNA Library Construction, Nuclease Protection Assays, Northern Blotting
High-throughput CompatibilityNot High-throughput Compatible (Manual)
No. of Reactions200 Preps
Purification Time1 hr.
Quantity100 mL
ScaleScaleable
Starting Material AmountUp to 1 g tissue, Up to 1 x 10^7 cells
Isolation TechnologyOrganic Extraction
Sample TypePlant Samples
TargetTotal RNA
Unit SizeEach
Contents & Storage
PureLink™ Plant RNA Reagent is supplied as a ready-to-use solution. Store at +4°C.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link