Calcium chloride dihydrate, extra pure, flakes for drying purposes, 77-80% CaCl2
Calcium chloride dihydrate, extra pure, flakes for drying purposes, 77-80% CaCl2
Calcium chloride dihydrate, extra pure, flakes for drying purposes, 77-80% CaCl2
Calcium chloride dihydrate, extra pure, flakes for drying purposes, 77-80% CaCl2
Thermo Scientific Chemicals

Calcium chloride dihydrate, extra pure, flakes for drying purposes, 77-80% CaCl2

CAS: 10035-04-8 | CaCl2·2H2O | 147.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
10 kg
Packaging:
View all
Plastic Bottle
Catalog number 123350025
Price (USD)
128.65
Special offer
Online exclusive
151.00
Save 22.35 (15%)
Each
-
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)
128.65
Special offer
Online exclusive
151.00
Save 22.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS10035-04-8
IUPAC Namecalcium dihydrate dichloride
Molecular FormulaCaCl2H4O2
InChI KeyLLSDKQJKOVVTOJ-UHFFFAOYSA-L
SMILESO.O.[Cl-].[Cl-].[Ca++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light grey
Appearance (Form)Flakes
Titration Complexometric77 to 80 % (CaCl2)
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0469510Certificate of AnalysisMay 17, 2025123350025, 123350100
A0471103Certificate of AnalysisMay 17, 2025123350025, 123350100
A0443331Certificate of AnalysisFeb 26, 2024123350025, 123350100
A0457989Certificate of AnalysisJan 25, 2024123350025, 123350100
A0394596Certificate of AnalysisDec 15, 2022123350025, 123350100
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection