Informational:Inquire about OEM or Commercial Supply version of this product here.
Invitrogen™

Platinum™ SuperFi™ DNA Polymerase

Catalog number: 12351050
Have Questions?
Invitrogen™

Platinum™ SuperFi™ DNA Polymerase

Catalog number: 12351050
Have Questions?
Catalog Number
12351050
Unit Size
500 units
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
100 units
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
5 x 500 units
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
12351050500 units
-
12351010100 units
-
123512505 x 500 units
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

Invitrogen™ Platinum™ SuperFi™ DNA Polymerase is a proofreading DNA polymerase that combines superior fidelity with trusted Platinum™ hot-start technology for the highest success in PCR. Featuring >300X Taq fidelity, Platinum SuperFi DNA Polymerase is ideally suited for cloning, mutagenesis, and other applications benefiting from supreme sequence accuracy.

Benefits of Platinum SuperFi DNA Polymerase include:
• Exceptional >300X Taq fidelity
• High specificity and increased yields with Platinum hot-start technology
• Robust amplification of difficult-to-amplify targets including those of sub-optimal purity or with ˃65% GC content
• Convenient workflow with room temperature reaction setup and 24 hr. bench top stability of pre-assembled reactions

Platinum SuperFi DNA Polymerase is engineered with a DNA-binding domain resulting in high processivity and increased resistance to PCR inhibitors. This feature also enables fast-cycling protocols and amplification of long targets. The Platinum hot-start technology is based on proprietary antibodies that inhibit enzyme activity until the initial PCR denaturation step, preventing non-specific amplification and primer degradation.

Applications of Platinum SuperFi DNA Polymerase
• High-fidelity PCR
• Cloning and sub-cloning
• Site-directed mutagenesis
• Amplification of GC-rich templates
• Template generation for sequencing
• High-throughput PCR
• Amplification of samples with suboptimal purity
• Long PCR
• Fast PCR

Using Platinum SuperFi DNA Polymerase
Annealing rules for Platinum SuperFi DNA Polymerase are different from many common DNA polymerases (such as Taq DNA polymerases). For optimal results, use the Tm calculator on our website: www.thermofisher.com/tmcalculator.

Platinum SuperFi DNA Polymerase is supplied with a separate vial of SuperFi™ GC Enhancer designed for difficult and GC-rich templates (˃65% GC).

Platinum hot-start technology efficiently inhibits DNA polymerase activity at ambient temperatures, allowing convenient reaction setup at room temperature. In addition, pre-assembled PCR reactions may be stored at room temperature for up to 24 hr. prior to the PCR.

More Platinum SuperFi DNA Polymerase products ›

For Research Use Only. Not for use in diagnostic procedures.

Specifications

GC-Rich PCR Performance
High
Polymerase
Platinum SuperFi DNA Polymerase
Reaction Speed
Fast
Shipping Condition
Wet or Dry Ice
For Use With (Application)
Hot-start PCR, High-fidelity PCR
Fidelity (vs. Taq)
300X
Hot Start
Built-In Hot Start
No. of Reactions
500 Reactions
Overhang
Blunt
Reaction Format
Separate Components
Size (Final Product)
20 kb or less

Contents & Storage

• Platinum SuperFi DNA Polymerase (250 μL at 2 U/μL)
• 5X SuperFi Buffer (6 x 1.25 mL)
• 5X SuperFi GC Enhancer (4 x 1.25 mL)

Store at –5°C to –30°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text