Invitrogen™

Platinum™ SuperFi™ DNA Polymerase

產品號碼: 12351250
Invitrogen™

Platinum™ SuperFi™ DNA Polymerase

產品號碼: 12351250
產品號碼
12351250
包裝規格
5 x 500 units
價格 (HKD)
價格 42,156.00
您的價格
供應狀況
***
數量
產品號碼
包裝規格
100 units
價格 (HKD)
價格 2,152.00
您的價格
供應狀況
***
數量
產品號碼
包裝規格
500 units
價格 (HKD)
價格 9,913.00
您的價格
供應狀況
***
數量
產品號碼包裝規格價格 (HKD)供應狀況數量
123512505 x 500 units
價格 42,156.00
您的價格
***
12351010100 units
價格 2,152.00
您的價格
***
12351050500 units
價格 9,913.00
您的價格
***
產品概覽
圖表
影片
建議
建議
文件
常見問題
引用資料與參考文獻
其他資訊
建議

Invitrogen™ Platinum™ SuperFi™ DNA Polymerase is a proofreading DNA polymerase that combines superior fidelity with trusted Platinum™ hot-start technology for the highest success in PCR. Featuring >300X Taq fidelity, Platinum SuperFi DNA Polymerase is ideally suited for cloning, mutagenesis, and other applications benefiting from supreme sequence accuracy.

Benefits of Platinum SuperFi DNA Polymerase include:
• Exceptional >300X Taq fidelity
• High specificity and increased yields with Platinum hot-start technology
• Robust amplification of difficult-to-amplify targets including those of sub-optimal purity or with ˃65% GC content
• Convenient workflow with room temperature reaction setup and 24 hr. bench top stability of pre-assembled reactions

Platinum SuperFi DNA Polymerase is engineered with a DNA-binding domain resulting in high processivity and increased resistance to PCR inhibitors. This feature also enables fast-cycling protocols and amplification of long targets. The Platinum hot-start technology is based on proprietary antibodies that inhibit enzyme activity until the initial PCR denaturation step, preventing non-specific amplification and primer degradation.

Applications of Platinum SuperFi DNA Polymerase
• High-fidelity PCR
• Cloning and sub-cloning
• Site-directed mutagenesis
• Amplification of GC-rich templates
• Template generation for sequencing
• High-throughput PCR
• Amplification of samples with suboptimal purity
• Long PCR
• Fast PCR

Using Platinum SuperFi DNA Polymerase
Annealing rules for Platinum SuperFi DNA Polymerase are different from many common DNA polymerases (such as Taq DNA polymerases). For optimal results, use the Tm calculator on our website: www.thermofisher.com/tmcalculator.

Platinum SuperFi DNA Polymerase is supplied with a separate vial of SuperFi™ GC Enhancer designed for difficult and GC-rich templates (˃65% GC).

Platinum hot-start technology efficiently inhibits DNA polymerase activity at ambient temperatures, allowing convenient reaction setup at room temperature. In addition, pre-assembled PCR reactions may be stored at room temperature for up to 24 hr. prior to the PCR.

More Platinum SuperFi DNA Polymerase products ›

For Research Use Only. Not for use in diagnostic procedures.

規格

GC-Rich PCR Performance
High
Polymerase
Platinum SuperFi DNA Polymerase
Reaction Speed
Fast
Shipping Condition
Wet or Dry Ice
For Use With (Application)
Hot-start PCR, High-fidelity PCR
Fidelity (vs. Taq)
300X
Hot Start
Built-In Hot Start
No. of Reactions
5 x 500 Reactions
Overhang
Blunt
Reaction Format
Separate Components
Size (Final Product)
20 kb or less

內容物與存放

5 boxes of Platinum SuperFi Master Mix, each containing:
• Platinum SuperFi DNA Polymerase (250 μL at 2 U/μL)
• 5X SuperFi Buffer (6 x 1.25 mL)
• 5X SuperFi GC Enhancer (4 x 1.25 mL)

Store at –5°C to –30°C.

圖表

文件與下載

證書

    常見問答集 (常見問題)

    引用資料與參考文獻

    Search citations by name, author, journal title or abstract text