Gibco™

CD Hybridoma AGT™ Medium

Catalog number: 12372001
Have Questions?
Gibco™

CD Hybridoma AGT™ Medium

Catalog number: 12372001
Have Questions?
Catalog Number
12372001
also known as 12372-001
Unit Size
100 L
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1 L
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
10 L
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
12372001
also known as 12372-001
100 L
-
123720251 L
-
1237201710 L
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Gibco™ CD Hybridoma Medium is an animal origin-free, chemically-defined, protein-free medium specifically developed for the ability to support the growth of hybridomas for antibody production. Gibco™ CD Hybridoma Medium features:
• Ability to support suspension and stationary hybridomas and myelomas
• Formulated without L-glutamine for stability
• Animal origin-free, protein-free and chemically defined formulation
• Convenient, easy-to-use Advanced Granulation Technology (AGT™) format

Ability to support suspension and stationary hybridomas and myelomas
Suitable for the culture of recombinant myeloma lines as well as traditional hybridomas, Gibco™ CD Hybridoma Medium provides high cell densities and IgG production. Cholesterol-dependent cultures such as NS0 require the addition of 250x Cholesterol Lipid Concentrate.

Formulated without L-glutamine for stability
Gibco™ CD Hybridoma Medium requires the addition of 8 mM L-glutamine or GlutaMAX™ supplement before use. GlutaMAX™ supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format.

Animal origin-free, protein-free and chemically defined formulation
Gibco™ CD Hybridoma Medium is animal origin-free, protein-free and chemically defined, allowing for easier purification of your protein of interest. Gibco™ chemically defined media contain no proteins, hydrolysates or components of unknown composition.

Convenient, easy-to-use Advanced Granulation Technology (AGT™) format
Our Advanced Granulation Technology™ (AGT™) format is an easy-to-use dry powder format. The granules dissolve instantly and pH-adjusts for fast media preparation. Complete reconstitution instructions are available in the product manual.

Product Use
For research use/further cell culture manufacturing. Not for use in diagnostic procedures. Customers using Gibco™ CD Hybridoma Medium in a manufacturing process, who have a submission with the FDA, may request a letter of authorization from us to reference our Type II Drug Master File (DMF).

cGMP Manufacturing and Quality System
Gibco™ CD Hybridoma Medium is manufactured at a cGMP compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

Specifications

Culture Environment
CO2
Cell Type
Myeloma Cells, Hybridomas
Classification
Animal Origin-free, Chemically-defined, Protein-free
Concentration
1 X
Form
Powder (AGT)
Product Type
Hybridoma Medium
Serum Level
Serum-free
Sterility
Non-sterile
With Additives
Sodium Pyruvate, Sodium Bicarbonate
Without Additives
No Glutamine, No HEPES, No Phenol Red
Endotoxin Level
Low
Glucose Concentration
3.66 g/L
Inorganic Salts
Calcium, Magnesium, Sodium Phosphate
Product Line
AGT™, Gibco™
Purity or Quality Grade
Certified
Quantity
100 L
Shipping Condition
Wet Ice

Contents & Storage

Storage conditions: 2-8°C, dark and dry
Shipping conditions: Gel packs
Shelf life: 24 months from date of manufacture

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text