Gibco™

CD 293 AGT™ Medium

Catalog number: 12529003
Have Questions?
Gibco™

CD 293 AGT™ Medium

Catalog number: 12529003
Have Questions?
Catalog Number
12529003
Unit Size
1 x 10kg
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
10 L
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1 L
Price (USD)
Availability
-
Quantity
Catalog Number
12529001
also known as 12529-001
Unit Size
100 L
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
125290031 x 10kg
-
1252901210 L
-
125290201 L
-
12529001
also known as 12529-001
100 L
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
CD 293 AGT™ is a chemically-defined, protein-free medium in a new easy-to-use granular format. CD 293 AGT™ is optimized for the growth of suspension cultures of 293 cells.
Advanced Granulation Technology = AGT
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Specifications

Culture Environment
CO2
Cell Line
293 (HEK)
Classification
Animal Origin-free
Culture Type
Suspension Cell Culture
Form
Powder (AGT)
Product Type
CD 293 Medium
Serum Level
Serum-free
Sterility
Non-sterile
With Additives
Low Glucose, Sodium Pyruvate, Sodium Bicarbonate, HEPES
Without Additives
No Glutamine
Endotoxin Level
Low
Format
Ropak Bucket
Glucose Concentration
4500 mg/L
Included Antibiotics
No antibiotics
Inorganic Salts
Calcium, Magnesium, Sodium Phosphate
Product Line
Gibco™
Purity or Quality Grade
Certified
Shipping Condition
Dry Ice

Contents & Storage

Store at 2 to 8°C in the dark.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text