Gibco™

CD 293 AGT™ Medium

Catalog number: 12529012
Have Questions?
Gibco™

CD 293 AGT™ Medium

Catalog number: 12529012
Have Questions?
Catalog Number
12529012
Unit Size
10 L
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1 L
Price (USD)
Availability
-
Quantity
Catalog Number
12529001
also known as 12529-001
Unit Size
100 L
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1 x 10kg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
1252901210 L
-
125290201 L
-
12529001
also known as 12529-001
100 L
-
125290031 x 10kg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
CD 293 AGT™ is a chemically-defined, protein-free medium in a new easy-to-use granular format. CD 293 AGT™ is optimized for the growth of suspension cultures of 293 cells.
Advanced Granulation Technology = AGT
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Specifications

Cell Line
293 (HEK)
Classification
Animal Origin-free, Chemically-defined, Protein-free, Serum-free
Culture Type
Suspension Cell Culture
Form
Powder (AGT)
Product Type
CD 293 Medium
Serum Level
Serum-free
With Additives
Phenol Red, Sodium Pyruvate, Sodium Bicarbonate
Without Additives
No Glutamine
Endotoxin Level
≤10 EU/mL, Low
Format
Securitainer Capped Vial
Included Antibiotics
No antibiotics
Product Line
Gibco™
Quantity
10 L
Reconstituted Volume
10 L
Shipping Condition
Wet Ice

Contents & Storage

Store in refrigerator (2–8°C) and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text