2-Mercaptoethanol, 99%, pure
2-Mercaptoethanol, 99%, pure
2-Mercaptoethanol, 99%, pure
2-Mercaptoethanol, 99%, pure
Thermo Scientific Chemicals

2-Mercaptoethanol, 99%, pure

CAS: 60-24-2 | C2H6OS | 78.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
10 mL
250 mL
2.5 L
Packaging:
Glass bottle
Catalog number 125470100
Price (USD)
41.65
Online Exclusive
46.40
Save 4.75 (10%)
Each
In stock
Add to cart
Quantity:
10 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
41.65
Online Exclusive
46.40
Save 4.75 (10%)
Each
Add to cart
Chemical Identifiers
CAS60-24-2
IUPAC Name2-sulfanylethan-1-ol
Molecular FormulaC2H6OS
InChI KeyDGVVWUTYPXICAM-UHFFFAOYSA-N
SMILESOCCS
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4990 to 1.5020 (20°C, 589 nm)
Infrared spectrumConforms
GC>=98.5 %
Water=<0.5 %
Appearance (Form)Liquid
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Los clientes que vieron este artículo también vieron



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0473783Certificate of AnalysisJun 26, 2025125470010, 125472500, 125470025, 125470100
A0465849Certificate of AnalysisSep 09, 2024125470010, 125472500, 125470025, 125470100
A0457747Certificate of AnalysisNov 23, 2023125470010, 125472500, 125470025, 125470100
A0435647Certificate of AnalysisMay 05, 2023125470010, 125470025, 125470100, 125472500
A0450736Certificate of AnalysisMay 02, 2023125470010, 125472500, 125470025, 125470100
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection