2-Mercaptoethanol
2-Mercaptoethanol
Gibco™

2-Mercaptoethanol

Gibco™ 2-Mercaptoethanol (also known as beta-mercaptoethanol or BME) is a potent reducing agent used in cell culture media to preventRead more
Have Questions?
Catalog NumberQuantity
2198502350 mL
Catalog number 21985023
Price (USD)
56.65
Online Exclusive
61.75
Save 5.10 (8%)
Each
-
Add to cart
Quantity:
50 mL
Recurring order eligible. Learn more »
Price (USD)
56.65
Online Exclusive
61.75
Save 5.10 (8%)
Each
Add to cart
Ask our AI about this Product
Gibco™ 2-Mercaptoethanol (also known as beta-mercaptoethanol or BME) is a potent reducing agent used in cell culture media to prevent toxic levels of oxygen radicals. 2-Mercaptoethanol is not stable in solution, so most protocols require daily supplementation. Gibco 2-Mercaptoethanol contains 2-mercaptoethanol at a concentration of 55 mM in Dulbecco's phosphate buffered saline (DPBS).

cGMP manufacturing and quality system
Gibco Reducing Agent is manufactured at a cGMP compliant facility, located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeMammalian Cells
Concentration55 mM
Quantity50 mL
Shelf Life36 Months
Shipping ConditionRoom Temperature
FormLiquid
Product TypeBME
pH6 to 8
Unit SizeEach
Contents & Storage
Storage conditions: 2°C to 8°C
Shipping conditions: Room temperature
Shelf life: 36 months from date of manufacture
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

해당 제품을 검색한 고객들이 함께 검색한 제품



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3177321Certificate of AnalysisJun 22, 202521985023
3177319Certificate of AnalysisMay 30, 202521985023
3118280Certificate of AnalysisMay 04, 202521985023
3075237Certificate of AnalysisApr 25, 202521985023
3075207Certificate of AnalysisMar 19, 202521985023
5 results displayed, search above for a specific certificate

Safety Data Sheets

Frequently asked questions (FAQs)

The 2-Mercaptoethanol must be added to the medium immediately before use because activity rapidly declines in diluted form. You can supplement enough medium needed for that day.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Citations & References (2)

Citations & References
Abstract
Delivery of proteins into living cells by reversible membrane permeabilization with streptolysin-O.
Authors:Walev I, Bhakdi SC, Hofmann F, Djonder N, Valeva A, Aktories K, Bhakdi S,
Journal:Proc Natl Acad Sci U S A
PubMed ID:11248053
'The pore-forming toxin streptolysin O (SLO) can be used to reversibly permeabilize adherent and nonadherent cells, allowing delivery of molecules with up to 100 kDa mass to the cytosol. Using FITC-labeled albumin, 10(5)-10(6) molecules were estimated to be entrapped per cell. Repair of toxin lesions depended on Ca(2+)-calmodulin and on ... More
Somatic coding mutations in human induced pluripotent stem cells.
Authors:Gore A, Li Z, Fung HL, Young JE, Agarwal S, Antosiewicz-Bourget J, Canto I, Giorgetti A, Israel MA, Kiskinis E, Lee JH, Loh YH, Manos PD, Montserrat N, Panopoulos AD, Ruiz S, Wilbert ML, Yu J, Kirkness EF, Izpisua Belmonte JC, Rossi DJ, Thomson JA, Eggan K, Daley GQ, Goldstein LS, Zhang K,
Journal:Nature
PubMed ID:21368825
Defined transcription factors can induce epigenetic reprogramming of adult mammalian cells into induced pluripotent stem cells. Although DNA factors are integrated during some reprogramming methods, it is unknown whether the genome remains unchanged at the single nucleotide level. Here we show that 22 human induced pluripotent stem (hiPS) cell lines ... More
2 total citations

다른 제품들



Share catalog number, name or link

1x1 image pixel for data collection