alpha-Methylstyrene, 99%, stabilized
alpha-Methylstyrene, 99%, stabilized
alpha-Methylstyrene, 99%, stabilized
alpha-Methylstyrene, 99%, stabilized
Thermo Scientific Chemicals

alpha-Methylstyrene, 99%, stabilized

CAS: 98-83-9 | C9H10 | 118.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 127711000
Price (USD)
43.65
Special offer
Online exclusive
51.40
Save 7.75 (15%)
Each
In stock
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
43.65
Special offer
Online exclusive
51.40
Save 7.75 (15%)
Each
Add to cart
Chemical Identifiers
CAS98-83-9
IUPAC Name(prop-1-en-2-yl)benzene
Molecular FormulaC9H10
InChI KeyXYLMUPLGERFSHI-UHFFFAOYSA-N
SMILESCC(=C)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to almost colorless
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.5360 to 1.5400 (20°C, 589 nm)
Stabilizer10 to 20 ppm tert-Butylcatechol
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0440224Certificate of AnalysisMay 19, 2025127710010, 127711000, 127710025
A0472343Certificate of AnalysisMay 17, 2025127710010, 127711000, 127710025
A0462602Certificate of AnalysisMay 09, 2024127710010, 127711000, 127710025
A0460543Certificate of AnalysisFeb 22, 2024127710010, 127711000, 127710025
A0454064Certificate of AnalysisAug 25, 2023127710010, 127711000, 127710025
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection