4-Nitrotoluene, 99%
4-Nitrotoluene, 99%
4-Nitrotoluene, 99%
Thermo Scientific Chemicals

4-Nitrotoluene, 99%

CAS: 99-99-0 | C7H7NO2 | 137.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
1 kg
Packaging:
Glass bottle
Catalog number 129050050
Price (EUR)
19,13
Online Exclusive
22,50
Save 3,37 (15%)
Each
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Recurring order eligible. Learn more »
Request bulk or custom format
Price (EUR)
19,13
Online Exclusive
22,50
Save 3,37 (15%)
Each
Add to cart
Chemical Identifiers
CAS99-99-0
IUPAC Name1-methyl-4-nitrobenzene
Molecular FormulaC7H7NO2
InChI KeyZPTVNYMJQHSSEA-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
GC>=98.5 %
Appearance (Form)Crystalline solid
Infrared spectrumConforms
Melting point51°C to 54°C
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0471533Certificate of AnalysisMay 17, 2025129050050, 129050010, 129051000
A0469561Certificate of AnalysisJan 30, 2025129050050, 129050010, 129051000
A0465684Certificate of AnalysisAug 23, 2024129050050, 129050010, 129051000
A0434348Certificate of AnalysisMar 18, 2024129050050, 129050010, 129051000
A0450104Certificate of AnalysisFeb 09, 2023129050050, 129050010, 129051000
5 results displayed, search above for a specific certificate

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Acute oral toxicity
Category 3
Acute dermal toxicity
Category 3
Acute Inhalation Toxicity - Dusts and Mists
Category 3
Specific target organ toxicity - (repeated exposure)
Category 2
Chronic aquatic toxicity
Category 2
Label Elements
Signal Word

Danger

Hazard Statements

H301 + H311 + H331 - Toxic if swallowed, in contact with skin or if inhaled

H373 - May cause damage to organs through prolonged or repeated exposure

H411 - Toxic to aquatic life with long lasting effects

Precautionary Statements

P280 - Wear protective gloves/protective clothing

P301 + P310 - IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician

P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

P311 - Call a POISON CENTER or doctor/physician

Share catalog number, name or link

1x1 image pixel for data collection