Phenylurea, 97%
Phenylurea, 97%
Phenylurea, 97%
Thermo Scientific Chemicals

Phenylurea, 97%

CAS: 64-10-8 | C7H8N2O | 136.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
Packaging:
Plastic bottle
Catalog number 131022500
Price (USD)
88.65
Special offer
Online exclusive
104.00
Save 15.35 (15%)
Each
Estimated availability date  Pending  
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
88.65
Special offer
Online exclusive
104.00
Save 15.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS64-10-8
IUPAC Namephenylurea
Molecular FormulaC7H8N2O
InChI KeyLUBJCRLGQSPQNN-UHFFFAOYSA-N
SMILESNC(=O)NC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Almost white to beige
Infrared spectrumConforms
HPLC>=96.0 %
Appearance (Form)Crystalline powder or crystals
Melting point143°C to 149°C
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0436140Certificate of AnalysisMar 04, 2024131022500, 131021000
A0398098Certificate of AnalysisDec 15, 2022131022500, 131021000
A0406746Certificate of AnalysisDec 13, 2022131022500, 131021000
A0443117Certificate of AnalysisJun 14, 2022131022500, 131021000
A0435097Certificate of AnalysisOct 05, 2021131022500, 131021000
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection