Informational:Informational:Save up to 31% on your order of cell culture products. View promotion details »
Gibco™

HBSS, calcium, magnesium, no phenol red

Catalog number: 14025126
Have Questions?
Gibco™

HBSS, calcium, magnesium, no phenol red

Catalog number: 14025126
Have Questions?
Catalog Number
14025126
Unit Size
6 x 1,000 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
500 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
10 x 500 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1,000 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
140251266 x 1,000 mL
-
14025092500 mL
-
1402513410 x 500 mL
-
140250761,000 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations with calcium and magnesium are generally used as transport media, or for reagent preparation.

We offer a variety of Gibco™ HBSS formulations for a range of cell culture applications.


This HBSS is modified as follows:
WithWithout
• Calcium• Phenol Red
• Magnesium• Sodium Pyruvate
• Glucose 


The complete formulation is available.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Specifications

Additive
No Sodium Pyruvate
Classification
Animal Origin-Free
Dilution
1 X
Product Line
Gibco™
Product Type
Hanks' Balanced Salt Solution (HBSS)
Quantity
6 x 1000 mL
Solution Type
Hank's Balanced Salt
For Use With (Application)
Mammalian Cell Culture
Green Features
Sustainable packaging
Osmolality
270 - 305 mOsm/kg
pH
6.7 - 7.8
Phenol Red Indicator
No Phenol Red
Shipping Condition
Room Temperature
Sterility
Sterile-filtered
Form
Liquid
Manufacturing Quality
cGMP-compliant under the ISO 13485 standard

Contents & Storage

Storage conditions: 15–30°C
Shipping conditions: Ambient
Shelf life: 36 months from date of manufacture

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text