Urea, 99.5%, for analysis
Urea, 99.5%, for analysis
Urea, 99.5%, for analysis
Urea, 99.5%, for analysis
Thermo Scientific Chemicals

Urea, 99.5%, for analysis

CAS: 57-13-6 | CH4N2O | 60.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
25 kg
1 kg
Packaging:
View all
Plastic drum
Catalog number 140750050
Price (USD)
168.65
Special offer
Online exclusive
Ends: 26-Sep-2025
208.00
Save 39.35 (19%)
Each
In stock
Add to cart
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)
168.65
Special offer
Online exclusive
Ends: 26-Sep-2025
208.00
Save 39.35 (19%)
Each
Add to cart
Chemical Identifiers
CAS57-13-6
IUPAC Nameurea
Molecular FormulaCH4N2O
InChI KeyXSQUKJJJFZCRTK-UHFFFAOYSA-N
SMILESNC(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point132°C to 135°C
Titration with HClO4>=99.4 %
Insoluble matter=<0.01 %
Residue after ignition=<0.01 %
Heavy metals (as Pb)=<10 ppm
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0471215Certificate of AnalysisMay 17, 2025140750250, 140750010, 140750050
A0465297Certificate of AnalysisAug 22, 2024140750250, 140750010, 140750050
A0452539Certificate of AnalysisApr 27, 2023140750250, 140750010, 140750050
A0400195Certificate of AnalysisDec 15, 2022140750250, 140750010, 140750050
A0408844Certificate of AnalysisDec 13, 2022140750250, 140750010, 140750050
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection