3,4-Dimethoxybenzylamine, 97%
3,4-Dimethoxybenzylamine, 97%
3,4-Dimethoxybenzylamine, 97%
3,4-Dimethoxybenzylamine, 97%
Thermo Scientific Chemicals

3,4-Dimethoxybenzylamine, 97%

CAS: 5763-61-1 | C9H13NO2 | 167.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 140831000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS5763-61-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
GC>=96.0 %
Appearance (Form)Liquid
Refractive index1.5530 to 1.5570 (20°C, 589 nm)
Infrared spectrumConforms
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0428533Certificate of AnalysisSep 25, 2023140831000, 140830250, 140830050
A0420761Certificate of AnalysisJul 16, 2020140831000, 140830250, 140830050
2 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection