Informational:Informational:Save up to 31% on your order of cell culture products. View promotion details »
Gibco™

HBSS, no calcium, no magnesium, no phenol red

Catalog number: 14175103
Have Questions?
Gibco™

HBSS, no calcium, no magnesium, no phenol red

Catalog number: 14175103
Have Questions?
Catalog Number
14175103
Unit Size
10 x 500 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
500 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1,000 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
6 x 1,000 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
1417510310 x 500 mL
-
14175095500 mL
-
141750791,000 mL
-
141751456 x 1,000 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation.

We offer a variety of Gibco™ HBSS formulations for a range of cell culture applications.


This HBSS is modified as follows:
WithWithout
• Glucose• Calcium
 • Magnesium
 • Phenol Red


The complete formulation is available.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Specifications

Additive
No Sodium Pyruvate
Color
Clear
Dilution
1 X
Product Line
Gibco™
Product Type
Hanks' Balanced Salt Solution (HBSS)
Quantity
10 x 500 mL
Solution Type
Hank's Balanced Salt
For Use With (Application)
Mammalian Cell Culture
Green Features
Sustainable packaging
Osmolality
270 - 295 mOsm/kg
pH
6.7 - 7.8
Phenol Red Indicator
No Phenol Red
Sterility
Sterile-filtered
Form
Liquid
Manufacturing Quality
cGMP-compliant under the ISO 13485 standard

Contents & Storage

Storage conditions: 15°C to 30°C
Shipping conditions: Room temperature
Shelf life: 36 months from date of manufacture

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text