1x1 image pixel for data collection
4-Nitroanisole, 99+%
4-Nitroanisole, 99+%
4-Nitroanisole, 99+%
4-Nitroanisole, 99+%
Thermo Scientific Chemicals

4-Nitroanisole, 99+%

CAS: 100-17-4 | C7H7NO3 | 153.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
5 g
Packaging:
Glass bottle
Catalog number 147022500
Price (USD)
85.50
Each
In stock
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
85.50
Each
Add to cart
Chemical Identifiers
CAS100-17-4
IUPAC Name1-methoxy-4-nitrobenzene
Molecular FormulaC7H7NO3
InChI KeyBNUHAJGCKIQFGE-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals
Melting point51°C to 53°C
Appearance (Color)Green to brown
Infrared spectrumConforms
GC>=99.0 %
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0458410Certificate of AnalysisJun 11, 2025147022500, 147020050
A0395422Certificate of AnalysisApr 04, 2024147022500, 147020050
A0460181Certificate of AnalysisFeb 14, 2024147022500, 147020050
A0458410Certificate of AnalysisDec 12, 2023147022500, 147020050
A0453121Certificate of AnalysisMay 16, 2023147022500, 147020050
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link