Nunc™ Lab-Tek™ II Chamber Slide™ System
Save 51% on plasticware essentials and stock your lab to culture with confidence. View promotion details >
Nunc™ Lab-Tek™ II Chamber Slide™ System
Thermo Scientific™

Nunc™ Lab-Tek™ II Chamber Slide™ System

Grow cells directly on slides with the Thermo Scientific™ Nunc™ Lab-Tek™ II Chamber Slide™ System.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWellsMaterialDescriptionUnit Size
1544531RS Glass1-well Chamber Slide w/ removable wellCase of 96
154453PK
also known as 154453
1RS Glass1-well Chamber Slide w/ removable wellPack of 16
1544612RS Glass2-well Chamber Slide w/ removable wellsCase of 96
154461PK2RS Glass2-well Chamber Slide w/ removable wellsPack of 16
1545264RS Glass4-well Chamber Slide w/ removable wellsCase of 96
154526PK4RS Glass4-well Chamber Slide w/ removable wellsPack of 16
1545348RS Glass8-well Chamber Slide w/ removable wellsCase of 96
154534PK8RS Glass8-well Chamber Slide w/ removable wellsPack of 16
Catalog number 154453
Price (USD)
1,112.65
Online Exclusive
1,164.00
Save 51.35 (4%)
Case of 96
Estimated availability date 21-Jul-2025
Add to cart
Wells:
1
Material:
RS Glass
Description:
1-well Chamber Slide w/ removable well
Unit Size:
Case of 96
Recurring order eligible. Learn more »
Price (USD)
1,112.65
Online Exclusive
1,164.00
Save 51.35 (4%)
Case of 96
Add to cart
Ask our AI about this Product

Grow, stain, and examine cells directly on the Thermo Scientific™ Nunc™ Lab-Tek™ II Chamber Slide™ which consists of a removable polystyrene media chamber attached to a standard glass slide. Slide surface is specially treated for adherent cell culture. Available in 1-, 2-, 4-, and 8-well formats.

Features:
  • Slide surface is specially treated with a unique RS wash to ensure consistent cell growth.
  • Tabbed polystyrene lid allows gas exchange with minimal evaporation.
  • Removal tool separates media chamber from glass slide.
  • Biocompatible adhesive hydrophobic well border is printed on slide.
  • Slide features a white frosted writing area.

Includes:

  • Chamber removal tool

Ordering Information:

  • Can be purchased in case of 96 or pack of 16.
  • Shipped sterile in convenient stackable incubation trays.
Warranty: 90 days
WARNING: Cancer – www.P65Warnings.ca.gov
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Closure ColorClear
Description1-well Chamber Slide w/ removable well
IncludesClear polystyrene lid with handles on the top and side for aseptic manipulations
Lid MaterialPolystyrene
No. per Pack16
Culture Area8.6 cm2
Dimensions (LxW)74 x 25 mm
MaterialRS Glass
SterilitySterile
Surface TreatmentCell Culture
Volume (Metric) Working2.0 to 4.5 mL
Wells1
Unit SizeCase of 96
ECM
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Frequently asked questions (FAQs)

Unfortunately, we do not have a recommended cell density for our LabTek Chamber Slides. However, you can refer to the following link, which provides recommended cell densities for our other cell culture products:
https://www.thermofisher.com/us/en/home/references/gibco-cell-culture-basics/cell-culture-protocols/cell-culture-useful-numbers.html

The Nunc Lab-Tek II Chamber Slide System products are not coated with any chemicals or reagents.

Please review the following key differences between the Nunc Lab-Tek Chamber Slide System and the Nunc Lab-Tek II Chamber Slide System:
- The Nunc Lab-Tek Chamber Slide System is available in Permanox plastic or glass and incorporates a silicone gasket.
- The Nunc Lab-Tek II Chamber Slide System is available in glass only and do not have a silicone gasket.
- The Nunc Lab-Tek II Chamber Slide System has a blue hydrophobic border and the upper structure is attached by an adhesive. Supplied with this product is a key and holder system which is used to remove the upper structure from the glass slide.
- The Nunc Lab-Tek Chamber Slide System is available in 1-, 2-, 4-, 8- or 16-well formats.
- The Nunc Lab-Tek II Chamber Slide System is available in 1-, 2-, 4- or 8-well formats.

The RS treatment is a wash step formulated to improve the consistency of the glass surface of the Nunc Lab-Tek II Chamber Slide System. RS glass treatment prepares the glass surface for culture after the hydrophobic border has been applied.
The term "RS treatment" refers to the glass washing procedure.

Lab-Tek products are compatible with all common fixatives, including paraformaldehyde. Acetone at 100% and some mixtures of acetone and alcohols are not compatible with the polystyrene upper well structure and will result in clouding or other effects of chemical attack. We encourage careful testing within planned operating conditions before working with high-value samples.

Other products to consider



Share catalog number, name or link

1x1 image pixel for data collection