4-Nitrophenol, 99%
4-Nitrophenol, 99%
4-Nitrophenol, 99%
Thermo Scientific Chemicals

4-Nitrophenol, 99%

CAS: 100-02-7 | C6H5NO3 | 139.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
250 g
1 kg
5 kg
Packaging:
View all
Glass Bottle
Catalog number 157050050
Price (USD)
39.10
Each
In stock
Add to cart
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
39.10
Each
Add to cart
Chemical Identifiers
CAS100-02-7
IUPAC Name4-nitrophenol
Molecular FormulaC6H5NO3
InChI KeyBTJIUGUIPKRLHP-UHFFFAOYSA-N
SMILESOC1=CC=C(C=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point112°C to 115°C
Infrared spectrumConforms
GC>=98.5 %
Appearance (Color)Yellow to brown
Appearance (Form)Crystalline powder or crystals and/or chunks
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0473539Certificate of AnalysisJun 17, 2025157052500, 157050050, 157050051, 157050010, 157051000
A0458108Certificate of AnalysisDec 07, 2023157052500, 157050050, 157050051, 157050010, 157051000
B0149948Certificate of AnalysisNov 14, 2023157052500, 157050050, 157050051, 157050010, 157051000
A0453503Certificate of AnalysisAug 25, 2023157052500, 157050050, 157050051, 157050010, 157051000
A0402504Certificate of AnalysisDec 15, 2022157052500, 157050050, 157050051, 157050010, 157051000
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection