1,2,4,5-Tetrachlorobenzene, 98%
1,2,4,5-Tetrachlorobenzene, 98%
1,2,4,5-Tetrachlorobenzene, 98%
1,2,4,5-Tetrachlorobenzene, 98%
Thermo Scientific Chemicals

1,2,4,5-Tetrachlorobenzene, 98%

CAS: 95-94-3 | C6H2Cl4 | 215.89 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
5 g
Packaging:
Glass bottle
Catalog number 157782500
Price (USD)
312.65
Special offer
Online exclusive
368.00
Save 55.35 (15%)
Each
Estimated availability date  Pending  
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
312.65
Special offer
Online exclusive
368.00
Save 55.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS95-94-3
IUPAC Name1,2,4,5-tetrachlorobenzene
Molecular FormulaC6H2Cl4
InChI KeyJHBKHLUZVFWLAG-UHFFFAOYSA-N
SMILESClC1=CC(Cl)=C(Cl)C=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=97.5 %
Appearance (Color)White to pink to beige to grey
Melting point139°C to 142°C
Appearance (Form)Powder or crystalline powder or crystals
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0472815Certificate of AnalysisMay 20, 2025157782500, 157780050
A0397972Certificate of AnalysisMay 17, 2025157782500, 157780050
A0431196Certificate of AnalysisMay 17, 2025157782500, 157780050
A0431196Certificate of AnalysisDec 11, 2023157782500, 157780050
A0397972Certificate of AnalysisAug 25, 2023157782500, 157780050
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link