3-Chlorotoluene, 97%
3-Chlorotoluene, 97%
3-Chlorotoluene, 97%
3-Chlorotoluene, 97%
Thermo Scientific Chemicals

3-Chlorotoluene, 97%

CAS: 108-41-8 | C7H7Cl | 126.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
250 mL
500 mL
Packaging:
Glass bottle
Catalog number 160452500
Price (USD)
85.65
Special offer
Online exclusive
100.00
Save 14.35 (14%)
Each
Estimated availability date  Pending  
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
85.65
Special offer
Online exclusive
100.00
Save 14.35 (14%)
Each
Add to cart
Chemical Identifiers
CAS108-41-8
IUPAC Name1-chloro-3-methylbenzene
Molecular FormulaC7H7Cl
InChI KeyOSOUNOBYRMOXQQ-UHFFFAOYSA-N
SMILESCC1=CC=CC(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=96.0 %
Refractive index1.5210 to 1.5230 (20°C, 589 nm)
Appearance (Form)Liquid
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0448272Certificate of AnalysisJun 09, 2025160455000, 160451000, 160452500
A0448272Certificate of AnalysisDec 08, 2022160455000, 160451000, 160452500
A0398786Certificate of AnalysisJan 15, 2021160455000, 160451000, 160452500
A0422974Certificate of AnalysisSep 29, 2020160451000, 160452500, 160455000
4 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection