Gibco™

FreeStyle™ MAX Reagent

Catalog number: 16447750
Have Questions?
Gibco™

FreeStyle™ MAX Reagent

Catalog number: 16447750
Have Questions?
Catalog Number
16447750
Unit Size
10 x 15 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
15 ml
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
1644775010 x 15 mL
-
164471001 mL
-
1644750015 ml
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
FreeStyle™ MAX Reagent is a proprietary, animal origin–free formulation for transfecting plasmid DNA into eukaryotic cells that can easily be scaled up to produce large amounts of recombinant proteins. FreeStyle™ MAX Reagent allows the highest expression levels and transfection rates with lowest cytotoxicity in bio-production applications, and is specifically formulated for use with:

• FreeStyle™ 293-F Cells (suspension human embryonal kidney cells, Cat. No. R790-07) in serum-free FreeStyle™ 293 Expression Medium (Cat. No. 12338-018)
• FreeStyle™ CHO-S Cells (suspension Chinese Hamster Ovary cells, Cat. No. R800-07) in serum-free FreeStyle™ CHO Expression Medium (Cat. No. 12651-014)
• DG44 Cells (DHFR suspension CHO cells) in CD DG44 Medium with 8 mM L-glutamine and 18 mL/L 10% Pluronic™ F-68 (Cat. No. A11000-01)

Product use
FreeStyle™ MAX Reagent is intended for use with the FreeStyle™ MAX 293 Expression System (Cat. No. K9000-10) and the FreeStyle™ MAX CHO Expression System (Cat. No. K9000-20).
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Classification
Animal Origin-Free
Cell Type
Established Cell Lines
Format
Tube
Sample Type
Plasmid DNA
Transfection Technique
Lipid-based Transfection
For Use With (Application)
Protein Expression
High-throughput Compatibility
Not High-throughput Compatible (Manual)
Product Line
Freestyle™
Product Type
Transfection Reagent
Quantity
10 x 15 mL
Serum Compatible
Yes
Shipping Condition
Wet Ice

Contents & Storage

Each kit contains 10 units. Each unit contains one vial of reagent. Store at +4°C. FreeStyle™ MAX is guaranteed stable for 6 months when stored properly.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text