Poly(methyl methacrylate), sec. stand., approx. M.W. 90,000, fine granules
Poly(methyl methacrylate), sec. stand., approx. M.W. 90,000, fine granules
Poly(methyl methacrylate), sec. stand., approx. M.W. 90,000, fine granules
Poly(methyl methacrylate), sec. stand., approx. M.W. 90,000, fine granules
Thermo Scientific Chemicals

Poly(methyl methacrylate), sec. stand., approx. M.W. 90,000, fine granules

CAS: 9011-14-7 | (C5H8O2)n | NaN g/mol
Have Questions?
Catalog NumberQuantityPackaging
17878025025 gGlass bottle
Catalog number 178780250
Price (USD)
110.65
Special offer
Online exclusive
130.00
Save 19.35 (15%)
Each
In stock
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
110.65
Special offer
Online exclusive
130.00
Save 19.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS9011-14-7
Molecular Formula(C5H8O2)n
SMILESCOC(=O)C(C)(-*)C-*
MDL NumberMFCD00134349
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<5 %
Infrared spectrumConforms
Particle size50 to 150 microns (Approximately)
Appearance (Form)Beads
Appearance (Color)White
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection