EDTA
EDTA
Thermo Scientific™

EDTA

Thermo Scientific Pierce EDTA is useful as a chelator of alkaline earth metals, as well as iron, copper and zincRead more
Have Questions?
Catalog NumberQuantity
17892500 g
Catalog number 17892
Price (USD)
116.00
Each
-
Add to cart
Quantity:
500 g
Recurring order eligible. Learn more »
Price (USD)
116.00
Each
Add to cart
Ask our AI about this Product
Thermo Scientific Pierce EDTA is useful as a chelator of alkaline earth metals, as well as iron, copper and zinc in a variety of laboratory methods.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
CAS6381-92-6
Chemical Name or MaterialEDTA
Molecular FormulaC10H14N2Na2O8·2H2O
Product TypeEDTA (ethylenediaminetetraacetic acid)
Purity≥99%
Quantity500 g
SynonymEDTA disodium salt dihydrate
FormPowder
Unit SizeEach
Contents & Storage
Store at room temperature.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Frequently asked questions (FAQs)

Can I use EDTA (Cat. No. 17892) in molecular biology? If not, can you provide an alternative?

EDTA (Cat. No. 17892) is not intended for molecular biology applications.

UltraPure Ethylenediaminetetraacetic Acid, Disodium Salt, Dihydrate (Na2EDTA·2H2O) (Cat. No. 15576028) is a potential alternative intended for molecular biology applications.

Do you offer any of the components of the Pierce protease, phosphatase and (protease and phosphatase) inhibitor tablets and mini tablets individually?

Yes, the following components of the Pierce protease, phosphatase and (protease and phosphatase) inhibitor tablets and mini tablets are offered separately:

- AEBSF (Cat. No. 78431)
- Aprotinin (Cat. No. 78432)
- Bestatin (Cat. No. 8433)
- E-64 (Cat. No. 78434)
- Leupeptin (Cat. No. 78435)
- Pepstatin A (Cat. No. 78436)
- EDTA (Cat. No. 17892).

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Kunden, die diesen Artikel ansahen, interessierten sich auch für



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
79422794Certificate of AnalysisJun 05, 202517892
ZJ4234201Certificate of AnalysisOct 15, 202417892
ZD4215681Certificate of AnalysisApr 17, 202417892
YC3657473Certificate of AnalysisMar 09, 202317892
XJ3468351ACertificate of AnalysisNov 24, 202217892
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection