Thermo Scientific™

Phenol:Chloroform + Tris Buffer

Catalog number: 17908
Have Questions?
Thermo Scientific™

Phenol:Chloroform + Tris Buffer

Catalog number: 17908
Have Questions?
Catalog Number
17908
Unit Size
100 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
400 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
17908100 mL
-
17909400 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Pierce Phenol-Chloroform is phenol supplied premixed with chloroform:isoamyl alcohol (25:24:1) and accompanying bottle of buffer to equilibrate at either pH 6.7 or pH 8.0.

Features of Phenol:Chloroform + Tris Buffer :

• Offers excellent, nuclease-free performance
• Phenol is supplied premixed with Chloroform:Isoamyl Alcohol (25:24:1)
• Supplied at pH 6.7 for special, lower pH DNA extractions; for most DNA applications, the alkaline buffer (included) should be added to produce a pH of 8.0
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Application)
DNA Extraction
Product Type
Phenol Chloroform and Tris Buffer
Quantity
100 mL
Form
Liquid

Contents & Storage

Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 2 to 8°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text