Invitrogen™

DNA Polymerase I, Large (Klenow) Fragment

Catalog number: 18012021
Have Questions?
Invitrogen™

DNA Polymerase I, Large (Klenow) Fragment

Catalog number: 18012021
Have Questions?
Catalog Number
18012021
Unit Size
100 units
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
500 units
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
18012021100 units
-
18012039500 units
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
DNA Polymerase I, Large (Klenow) Fragment is a DNA polymerase enzyme that lacks the 5' to 3' exonuclease activity of intact DNA Polymerase I, but does exhibit the 5' to 3' DNA polymerase and 3' to 5' exonuclease activities.

Applications:
Fill-in of 5´ overhangs (1). Synthesis of probes by random primers labeling method (2). Sequencing single- and double-stranded DNA (3). Site-directed mutagenesis.

Source:
Purified from E. coli expressing the Klenow fragment on a plasmid.

Performance and Quality Testing:
SDS-PAGE purity; single-stranded and double-stranded endodeoxyribonuclease and self-priming assays; performance evaluated in fill-in reaction.

Unit Definition:
One unit incorporates 10 nmol of total deoxyribonucleotide into acid-precipitable material in 30 min. at 37°C using a template•primer.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Description
DNA Polymerase
Polymerase
DNA Polymerase I
Exonuclease Activity
3' to 5'
Quantity
100 Units
Shipping Condition
Approved for shipment on Wet or Dry Ice
Hot Start
No

Contents & Storage

DNA Polymerase I, Large (Klenow) Fragment is supplied with a vial of 10X REact™ 2 Buffer [500 mM Tris-HCl (pH 8.0), 100 mM
MgCl2 , 500 mM NaCl], vial of dilution buffer. Store at -20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text